Order Entry
Canada
ContactUsLinkComponent
Human;Mouse;Rat Recombinant BDNF (from E. coli)
Human;Mouse;Rat Recombinant BDNF (from E. coli)
Supplier:  Peprotech
Certificates
Human;Mouse;Rat Recombinant BDNF (from E. coli)
Supplier:  Peprotech
Certificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
    Rat
    Mouse
  • Storage conditions:
    Store at –20 °C prior to reconstitution.
  • Endotoxin content:
    >0.1 ng/μg
  • Biological activity:
    Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Form:
    Lyophilized
  • Gene ID:
    627 P23560
  • Reconstitution instructions:
    Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Protease-free:
    Y
  • Protein synonyms:
    Brain-Derived Neurotrophic Factor|Abrineurin
  • Protein/peptide name:
    BDNF
  • Purity:
    0.98
  • Molecular weight:
    27 kDa
  • Sequence:
    MHSDPARRGE LSVCDSISEW VTAADKKTAV DMSGGTVTVL EKVPVSKGQL KQYFYETKCN PMGYTKEGCR GIDKRHWNSQ CRTTQSYVRA LTMDSKKRIG WRFIRIDTSC VCTLTIKRGR
  • Endotoxin level:
    Low
  • Formulation:
    Formulation can vary. Please refer to product CoA for information.
  • Shipping temperature:
    Ambient

Specifications

Frequently Bought Together