Rat Recombinant IL-7 (from E. coli)
IL-7 is a hematopoietic growth factor that primarily affects early B and T cells. Produced by thymic stromal cells, spleen cells and keratinocytes, IL-7 can also co-stimulate the proliferation of mature T cells in combination with other factors, such as ConA and IL-2. Human and murine IL-7 is cross-species reactive. Recombinant Rat IL-7 is a 15.0 kDa protein containing 130 amino acid residues.
: For research use only. Not for use in diagnostic or therapeutic procedures.
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Rat
- Storage conditions:Store at –20 °C prior to reconstitution.
- Endotoxin content:>0.1 ng/μg
- Biological activity:Biological Activity tests can vary. Please refer to the Product CoA for information.
- Form:Lyophilized
- Gene ID:25647 P56478
- Reconstitution instructions:Reconstitution instructions can vary. Please refer to the product CoA for info.
- Protease-free:Y
- Protein synonyms:Lymphopoietin 1(LP-1)|Interleukin-7|pre-B-cell factor
- Protein/peptide name:IL-7
- Purity:0.98
- Molecular weight:15 kDa
- Sequence:MDCHIKDKDG KAFGSVLMIS INQLDKMTGT DSDCPNNEPN FFKKHLCDDT KEAAFLNRAA RKLRQFLKMN ISEEFNDHLL RVSDGTQTLV NCTSKEEKTI KEQKKNDPCF LKRLLREIKT CWNKILKGSI
- Endotoxin level:Low
- Formulation:Formulation can vary. Please refer to product CoA for information.
- Shipping temperature:Ambient
Frequently Bought Together