Human Recombinant MIP-3beta (from E. coli)
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
MIP-3β is a CC chemokine, expressed in the thymus, lymph nodes, and in activated bone marrow stromal cells that signals through the CCR7 receptor. MIP-3β is a chemoattractant for T and B lymphocytes, and myeloid progenitor cells. Human MIP-3β is active on murine cells. Recombinant Human MIP-3β (CCL19) is an 8.8 kDa protein containing 77 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications
Specifications
- Size:5 μg
- Origin:US
- Protease-free:Y
- Form:Lyophilized
- Protein/peptide name:MIP-3beta
- Protein synonyms:SCYA19|ELC|Macrophage Inflammatory Protein-3β|Exodus-3
- Protein Function:Cytokines and Growth Factors
- Purity:0.98
- Endotoxin level:Low
- Sequence:GTNDAEDCCL SVTQKPIPGY IVRNFHYLLI KDGCRVPAVV FTTLRGRQLC APPDQPWVER IIQRLQRTSA KMKRRSS
- Source:E. coli
- Biological activity:Biological Activity tests can vary. Please refer to the Product CoA for information.
- Molecular weight:8.8 kDa
- Shipping temperature:Ambient
- Cat. No.:10780-526
- Reconstitution instructions:Reconstitution instructions can vary. Please refer to the product CoA for info.
- Protein/peptide type:Recombinant
- Endotoxin content:>0.1 ng/μg
- Storage conditions:Store at –20 °C prior to reconstitution.
- Gene ID:6363 Q99731
- Formulation:Formulation can vary. Please refer to product CoA for information.
- Conjugation:Unconjugated
- Species:Human
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



