Mouse Recombinant Eotaxin (from E. coli)
Eotaxin is a CC chemokine that signals through the CCR3 receptor. It is produced by IFN-γ-stimulated endothelial cells and TNF-activated monocytes. Eotaxin selectively chemoattracts eosinophils and, along with Eotaxin-2 and Eotaxin-3, plays a key role in the regulation of eosinophil recruitment in the asthmatic lung and in allergic reactions. Recombinant Murine Eotaxin (CCL11) is an 8.4 kDa protein containing 74 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines.
: For research use only. Not for use in diagnostic or therapeutic procedures.
Eotaxin is a CC chemokine that signals through the CCR3 receptor. It is produced by IFN-γ-stimulated endothelial cells and TNF-activated monocytes. Eotaxin selectively chemoattracts eosinophils and, along with Eotaxin-2 and Eotaxin-3, plays a key role in the regulation of eosinophil recruitment in the asthmatic lung and in allergic reactions. Recombinant Murine Eotaxin (CCL11) is an 8.4 kDa protein containing 74 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines.
: For research use only. Not for use in diagnostic or therapeutic procedures.
- Catalog No:
- 10780-224
- 10780-226
- 10780-228
- 10780-230
- Protein/peptide name:
- Eotaxin
- Eotaxin
- Eotaxin
- Eotaxin
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein Function:
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Species:
- Mouse
- Mouse
- Mouse
- Mouse
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- 20292 P48298
- 20292 P48298
- 20292 P48298
- 20292 P48298
- Sequence:
- HPGSIPTSCC FIMTSKKIPN TLLKSYKRIT NNRCTLKAIV FKTRLGKEIC ADPKKKWVQD ATKHLDQKLQ TPKP
- HPGSIPTSCC FIMTSKKIPN TLLKSYKRIT NNRCTLKAIV FKTRLGKEIC ADPKKKWVQD ATKHLDQKLQ TPKP
- HPGSIPTSCC FIMTSKKIPN TLLKSYKRIT NNRCTLKAIV FKTRLGKEIC ADPKKKWVQD ATKHLDQKLQ TPKP
- HPGSIPTSCC FIMTSKKIPN TLLKSYKRIT NNRCTLKAIV FKTRLGKEIC ADPKKKWVQD ATKHLDQKLQ TPKP
- Protease-free:
- Y
- Y
- Y
- Y
- Endotoxin level:
- Low
- Low
- Low
- Low
- Endotoxin content:
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- Biological activity:
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Purity:
- 0.98
- 0.98
- 0.98
- 0.98
- Form:
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Formulation:
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Molecular weight:
- 8.4 kDa
- 8.4 kDa
- 8.4 kDa
- 8.4 kDa
- Reconstitution instructions:
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Storage conditions:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping temperature:
- Ambient
- Ambient
- Ambient
- Ambient
Product Family Options
- SizeOrigin
- : 250-01-5UG
Specifications
Cat. No.10780-224Protein/peptide nameEotaxinProtein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID20292 P48298SequenceHPGSIPTSCC FIMTSKKIPN TLLKSYKRIT NNRCTLKAIV FKTRLGKEIC ADPKKKWVQD ATKHLDQKLQ TPKPProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight8.4 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - : 250-01-20UG
Specifications
Cat. No.10780-226Protein/peptide nameEotaxinProtein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID20292 P48298SequenceHPGSIPTSCC FIMTSKKIPN TLLKSYKRIT NNRCTLKAIV FKTRLGKEIC ADPKKKWVQD ATKHLDQKLQ TPKPProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight8.4 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - : 250-01-50UG
Specifications
Cat. No.10780-228Protein/peptide nameEotaxinProtein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID20292 P48298SequenceHPGSIPTSCC FIMTSKKIPN TLLKSYKRIT NNRCTLKAIV FKTRLGKEIC ADPKKKWVQD ATKHLDQKLQ TPKPProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight8.4 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - : 250-01-100UG
Specifications
Cat. No.10780-230Protein/peptide nameEotaxinProtein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID20292 P48298SequenceHPGSIPTSCC FIMTSKKIPN TLLKSYKRIT NNRCTLKAIV FKTRLGKEIC ADPKKKWVQD ATKHLDQKLQ TPKPProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight8.4 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient



