Order Entry
Canada
Orders LinkContactUsLinkComponent
Human Recombinant WISP-1 (from <i>E. coli</i>)

Human Recombinant WISP-1 (from E. coli)

Supplier:   Peprotech
Human Recombinant WISP-1 (from <i>E. coli</i>)
Supplier:  Peprotech
Certificates

6 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

WISP-1 is a member of the CCN family of secreted, cysteine-rich regulatory proteins. It is expressed in the heart, kidney, lung, pancreas, placenta, ovary, small intestine and spleen. WISP-1 is a β-catenin-regulated protein that can contribute to tumorigenesis, and has also been shown to play a role in bone development and fracture repair. Recombinant Human WISP-1 is a 38.1 kDa protein containing 345 amino acid residues. It is composed of four distinct structural domains (modules): the IGF binding protein (IGFBP) domain, the von Willebrand Factor C (VWFC) domain, the thrombospondin type-1 repeat (TSP type-1) domain, and a C-terminal cysteine knot-like (CTCK) domain.

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 10779-162
  • 10779-164
  • 10779-166
  • 10779-168
  • 10779-170
  • 10779-172
  • Protein/peptide name:
  • WISP-1
  • WISP-1
  • WISP-1
  • WISP-1
  • WISP-1
  • WISP-1
  • Protein synonyms:
  • CCN4|Wnt-1 Inducible Signaling Pathway protein-1|Wnt-1-induced secreted protein
  • CCN4|Wnt-1 Inducible Signaling Pathway protein-1|Wnt-1-induced secreted protein
  • CCN4|Wnt-1 Inducible Signaling Pathway protein-1|Wnt-1-induced secreted protein
  • CCN4|Wnt-1 Inducible Signaling Pathway protein-1|Wnt-1-induced secreted protein
  • CCN4|Wnt-1 Inducible Signaling Pathway protein-1|Wnt-1-induced secreted protein
  • CCN4|Wnt-1 Inducible Signaling Pathway protein-1|Wnt-1-induced secreted protein
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein Function:
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Species:
  • Human
  • Human
  • Human
  • Human
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • 8840 O95388
  • 8840 O95388
  • 8840 O95388
  • 8840 O95388
  • 8840 O95388
  • 8840 O95388
  • Sequence:
  • TALSPAPTTM DFTPAPLEDT SSRPQFCKWP CECPPSPPRC PLGVSLITDG CECCKMCAQQ LGDNCTEAAI CDPHRGLYCD YSGDRPRYAI GVCAQVVGVG CVLDGVRYNN GQSFQPNCKY NCTCIDGAVG CTPLCLRVRP PRLWCPHPRR VSIPGHCCEQ WVCEDDAKRP RKTAPRDTGA FDAVGEVEAW HRNCIAYTSP WSPCSTSCGL GVSTRISNVN AQCWPEQESR LCNLRPCDVD IHTLIKAGKK CLAVYQPEAS MNFTLAGCIS TRSYQPKYCG VCMDNRCCIP YKSKTIDVSF QCPDGLGFSR QVLWINACFC NLSCRNPNDI FADLESYPDF SEIAN
  • TALSPAPTTM DFTPAPLEDT SSRPQFCKWP CECPPSPPRC PLGVSLITDG CECCKMCAQQ LGDNCTEAAI CDPHRGLYCD YSGDRPRYAI GVCAQVVGVG CVLDGVRYNN GQSFQPNCKY NCTCIDGAVG CTPLCLRVRP PRLWCPHPRR VSIPGHCCEQ WVCEDDAKRP RKTAPRDTGA FDAVGEVEAW HRNCIAYTSP WSPCSTSCGL GVSTRISNVN AQCWPEQESR LCNLRPCDVD IHTLIKAGKK CLAVYQPEAS MNFTLAGCIS TRSYQPKYCG VCMDNRCCIP YKSKTIDVSF QCPDGLGFSR QVLWINACFC NLSCRNPNDI FADLESYPDF SEIAN
  • TALSPAPTTM DFTPAPLEDT SSRPQFCKWP CECPPSPPRC PLGVSLITDG CECCKMCAQQ LGDNCTEAAI CDPHRGLYCD YSGDRPRYAI GVCAQVVGVG CVLDGVRYNN GQSFQPNCKY NCTCIDGAVG CTPLCLRVRP PRLWCPHPRR VSIPGHCCEQ WVCEDDAKRP RKTAPRDTGA FDAVGEVEAW HRNCIAYTSP WSPCSTSCGL GVSTRISNVN AQCWPEQESR LCNLRPCDVD IHTLIKAGKK CLAVYQPEAS MNFTLAGCIS TRSYQPKYCG VCMDNRCCIP YKSKTIDVSF QCPDGLGFSR QVLWINACFC NLSCRNPNDI FADLESYPDF SEIAN
  • TALSPAPTTM DFTPAPLEDT SSRPQFCKWP CECPPSPPRC PLGVSLITDG CECCKMCAQQ LGDNCTEAAI CDPHRGLYCD YSGDRPRYAI GVCAQVVGVG CVLDGVRYNN GQSFQPNCKY NCTCIDGAVG CTPLCLRVRP PRLWCPHPRR VSIPGHCCEQ WVCEDDAKRP RKTAPRDTGA FDAVGEVEAW HRNCIAYTSP WSPCSTSCGL GVSTRISNVN AQCWPEQESR LCNLRPCDVD IHTLIKAGKK CLAVYQPEAS MNFTLAGCIS TRSYQPKYCG VCMDNRCCIP YKSKTIDVSF QCPDGLGFSR QVLWINACFC NLSCRNPNDI FADLESYPDF SEIAN
  • TALSPAPTTM DFTPAPLEDT SSRPQFCKWP CECPPSPPRC PLGVSLITDG CECCKMCAQQ LGDNCTEAAI CDPHRGLYCD YSGDRPRYAI GVCAQVVGVG CVLDGVRYNN GQSFQPNCKY NCTCIDGAVG CTPLCLRVRP PRLWCPHPRR VSIPGHCCEQ WVCEDDAKRP RKTAPRDTGA FDAVGEVEAW HRNCIAYTSP WSPCSTSCGL GVSTRISNVN AQCWPEQESR LCNLRPCDVD IHTLIKAGKK CLAVYQPEAS MNFTLAGCIS TRSYQPKYCG VCMDNRCCIP YKSKTIDVSF QCPDGLGFSR QVLWINACFC NLSCRNPNDI FADLESYPDF SEIAN
  • TALSPAPTTM DFTPAPLEDT SSRPQFCKWP CECPPSPPRC PLGVSLITDG CECCKMCAQQ LGDNCTEAAI CDPHRGLYCD YSGDRPRYAI GVCAQVVGVG CVLDGVRYNN GQSFQPNCKY NCTCIDGAVG CTPLCLRVRP PRLWCPHPRR VSIPGHCCEQ WVCEDDAKRP RKTAPRDTGA FDAVGEVEAW HRNCIAYTSP WSPCSTSCGL GVSTRISNVN AQCWPEQESR LCNLRPCDVD IHTLIKAGKK CLAVYQPEAS MNFTLAGCIS TRSYQPKYCG VCMDNRCCIP YKSKTIDVSF QCPDGLGFSR QVLWINACFC NLSCRNPNDI FADLESYPDF SEIAN
  • Protease-free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Y
  • Endotoxin level:
  • Low
  • Low
  • Low
  • Low
  • Low
  • Low
  • Endotoxin content:
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • Biological activity:
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Purity:
  • 0.98
  • 0.98
  • 0.98
  • 0.98
  • 0.98
  • 0.98
  • Form:
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Formulation:
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Molecular weight:
  • 38.1 kDa
  • 38.1 kDa
  • 38.1 kDa
  • 38.1 kDa
  • 38.1 kDa
  • 38.1 kDa
  • Reconstitution instructions:
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Storage conditions:
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Shipping temperature:
  • Ambient
  • Ambient
  • Ambient
  • Ambient
  • Ambient
  • Ambient

Specifications

Product Family Options

Product Information

  • Size
    Origin
    Availability
    Price
  • 5 μg
    US
    Log in for availability
    Log in for availability
    Each (5µG)
    Each (5µG) - $241.16
    $241.16
    Each (5µG)
       $241.16
    Minimum order quantity is 1

    Supplier Number: 120-18-5UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10779-162
    Protein/peptide name
    WISP-1
    Protein synonyms
    CCN4|Wnt-1 Inducible Signaling Pathway protein-1|Wnt-1-induced secreted protein
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    8840 O95388
    Sequence
    TALSPAPTTM DFTPAPLEDT SSRPQFCKWP CECPPSPPRC PLGVSLITDG CECCKMCAQQ LGDNCTEAAI CDPHRGLYCD YSGDRPRYAI GVCAQVVGVG CVLDGVRYNN GQSFQPNCKY NCTCIDGAVG CTPLCLRVRP PRLWCPHPRR VSIPGHCCEQ WVCEDDAKRP RKTAPRDTGA FDAVGEVEAW HRNCIAYTSP WSPCSTSCGL GVSTRISNVN AQCWPEQESR LCNLRPCDVD IHTLIKAGKK CLAVYQPEAS MNFTLAGCIS TRSYQPKYCG VCMDNRCCIP YKSKTIDVSF QCPDGLGFSR QVLWINACFC NLSCRNPNDI FADLESYPDF SEIAN
    Protease-free
    Y
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    38.1 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient
  • 20 μg
    US
    Log in for availability
    Log in for availability
    Each (20µG)
    Each (20µG) - $588.28
    $588.28
    Each (20µG)
       $588.28
    Minimum order quantity is 1

    Supplier Number: 120-18-20UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10779-164
    Protein/peptide name
    WISP-1
    Protein synonyms
    CCN4|Wnt-1 Inducible Signaling Pathway protein-1|Wnt-1-induced secreted protein
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    8840 O95388
    Sequence
    TALSPAPTTM DFTPAPLEDT SSRPQFCKWP CECPPSPPRC PLGVSLITDG CECCKMCAQQ LGDNCTEAAI CDPHRGLYCD YSGDRPRYAI GVCAQVVGVG CVLDGVRYNN GQSFQPNCKY NCTCIDGAVG CTPLCLRVRP PRLWCPHPRR VSIPGHCCEQ WVCEDDAKRP RKTAPRDTGA FDAVGEVEAW HRNCIAYTSP WSPCSTSCGL GVSTRISNVN AQCWPEQESR LCNLRPCDVD IHTLIKAGKK CLAVYQPEAS MNFTLAGCIS TRSYQPKYCG VCMDNRCCIP YKSKTIDVSF QCPDGLGFSR QVLWINACFC NLSCRNPNDI FADLESYPDF SEIAN
    Protease-free
    Y
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    38.1 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient
  • 100 μg
    US
    Log in for availability
    Log in for availability
    Each (100µG)
    Each (100µG) - $2,404.59
    $2,404.59
    Each (100µG)
       $2,404.59
    Minimum order quantity is 1

    Supplier Number: 120-18-100UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10779-166
    Protein/peptide name
    WISP-1
    Protein synonyms
    CCN4|Wnt-1 Inducible Signaling Pathway protein-1|Wnt-1-induced secreted protein
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    8840 O95388
    Sequence
    TALSPAPTTM DFTPAPLEDT SSRPQFCKWP CECPPSPPRC PLGVSLITDG CECCKMCAQQ LGDNCTEAAI CDPHRGLYCD YSGDRPRYAI GVCAQVVGVG CVLDGVRYNN GQSFQPNCKY NCTCIDGAVG CTPLCLRVRP PRLWCPHPRR VSIPGHCCEQ WVCEDDAKRP RKTAPRDTGA FDAVGEVEAW HRNCIAYTSP WSPCSTSCGL GVSTRISNVN AQCWPEQESR LCNLRPCDVD IHTLIKAGKK CLAVYQPEAS MNFTLAGCIS TRSYQPKYCG VCMDNRCCIP YKSKTIDVSF QCPDGLGFSR QVLWINACFC NLSCRNPNDI FADLESYPDF SEIAN
    Protease-free
    Y
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    38.1 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient
  • 250 μg
    US
    Log in for availability
    Log in for availability
    Each (250µG)
    Each (250µG) - $3,621.73
    $3,621.73
    Each (250µG)
       $3,621.73
    Minimum order quantity is 1

    Supplier Number: 120-18-250UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10779-168
    Protein/peptide name
    WISP-1
    Protein synonyms
    CCN4|Wnt-1 Inducible Signaling Pathway protein-1|Wnt-1-induced secreted protein
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    8840 O95388
    Sequence
    TALSPAPTTM DFTPAPLEDT SSRPQFCKWP CECPPSPPRC PLGVSLITDG CECCKMCAQQ LGDNCTEAAI CDPHRGLYCD YSGDRPRYAI GVCAQVVGVG CVLDGVRYNN GQSFQPNCKY NCTCIDGAVG CTPLCLRVRP PRLWCPHPRR VSIPGHCCEQ WVCEDDAKRP RKTAPRDTGA FDAVGEVEAW HRNCIAYTSP WSPCSTSCGL GVSTRISNVN AQCWPEQESR LCNLRPCDVD IHTLIKAGKK CLAVYQPEAS MNFTLAGCIS TRSYQPKYCG VCMDNRCCIP YKSKTIDVSF QCPDGLGFSR QVLWINACFC NLSCRNPNDI FADLESYPDF SEIAN
    Protease-free
    Y
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    38.1 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient
  • 500 μg
    US
    Log in for availability
    Log in for availability
    Each (500µG)
    Each (500µG) - $6,217.16
    $6,217.16
    Each (500µG)
       $6,217.16
    Minimum order quantity is 1

    Supplier Number: 120-18-500UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10779-170
    Protein/peptide name
    WISP-1
    Protein synonyms
    CCN4|Wnt-1 Inducible Signaling Pathway protein-1|Wnt-1-induced secreted protein
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    8840 O95388
    Sequence
    TALSPAPTTM DFTPAPLEDT SSRPQFCKWP CECPPSPPRC PLGVSLITDG CECCKMCAQQ LGDNCTEAAI CDPHRGLYCD YSGDRPRYAI GVCAQVVGVG CVLDGVRYNN GQSFQPNCKY NCTCIDGAVG CTPLCLRVRP PRLWCPHPRR VSIPGHCCEQ WVCEDDAKRP RKTAPRDTGA FDAVGEVEAW HRNCIAYTSP WSPCSTSCGL GVSTRISNVN AQCWPEQESR LCNLRPCDVD IHTLIKAGKK CLAVYQPEAS MNFTLAGCIS TRSYQPKYCG VCMDNRCCIP YKSKTIDVSF QCPDGLGFSR QVLWINACFC NLSCRNPNDI FADLESYPDF SEIAN
    Protease-free
    Y
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    38.1 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient
  • 1 mg
    US
    Log in for availability
    Log in for availability
    Each (1mg)
    Each (1mg) - $10,346.91
    $10,346.91
    Each (1mg)
       $10,346.91
    Minimum order quantity is 1

    Supplier Number: 120-18-01M
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10779-172
    Protein/peptide name
    WISP-1
    Protein synonyms
    CCN4|Wnt-1 Inducible Signaling Pathway protein-1|Wnt-1-induced secreted protein
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    8840 O95388
    Sequence
    TALSPAPTTM DFTPAPLEDT SSRPQFCKWP CECPPSPPRC PLGVSLITDG CECCKMCAQQ LGDNCTEAAI CDPHRGLYCD YSGDRPRYAI GVCAQVVGVG CVLDGVRYNN GQSFQPNCKY NCTCIDGAVG CTPLCLRVRP PRLWCPHPRR VSIPGHCCEQ WVCEDDAKRP RKTAPRDTGA FDAVGEVEAW HRNCIAYTSP WSPCSTSCGL GVSTRISNVN AQCWPEQESR LCNLRPCDVD IHTLIKAGKK CLAVYQPEAS MNFTLAGCIS TRSYQPKYCG VCMDNRCCIP YKSKTIDVSF QCPDGLGFSR QVLWINACFC NLSCRNPNDI FADLESYPDF SEIAN
    Protease-free
    Y
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    38.1 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.