Human Recombinant GMF-beta (from E. coli)
2 μg, US
Catalog # 10774-832
Supplier: Peprotech
Supplier #: 450-37-2UG
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
GMF-β is a brain-specific protein that belongs to the actin-binding proteins ADF structural family. GMF-β appears to play a role in the differentiation, maintenance, and regeneration of the nervous system. It also supports the progression of certain auto-immune diseases, possibly through its ability to induce the production and secretion of various pro-inflammatory cytokines. Recombinant Human GMF-β is a 16.5 kDa globular protein containing 141 amino acid residues, including one intramolecular disulfide bond.
: For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications
Specifications
- Size:2 μg
- Origin:US
- Protease-free:Y
- Form:Lyophilized
- Protein/peptide name:GMF-beta
- Protein synonyms:Glia Maturation Factor beta|GMFB
- Protein Function:Cytokines and Growth Factors
- Purity:0.98
- Endotoxin level:Low
- Sequence:SESLVVCDVA EDLVEKLRKF RFRKETNNAA IIMKIDKDKR LVVLDEELEG ISPDELKDEL PERQPRFIVY SYKYQHDDGR VSYPLCFIFS SPVGCKPEQQ MMYAGSKNKL VQTAELTKVF EIRNTEDLTE EWLREKLGFF H
- Source:E. coli
- Biological activity:Biological Activity tests can vary. Please refer to the Product CoA for information.
- Molecular weight:16.5 kDa
- Shipping temperature:Ambient
- Cat. No.:10774-832
- Reconstitution instructions:Reconstitution instructions can vary. Please refer to the product CoA for info.
- Protein/peptide type:Recombinant
- Endotoxin content:>0.1 ng/μg
- Storage conditions:Store at –20 °C prior to reconstitution.
- Gene ID:2764 P60983
- Formulation:Formulation can vary. Please refer to product CoA for information.
- Conjugation:Unconjugated
- Species:Human



