BAFF Receptor (BAFFR), a member of the TNFR superfamily, is highly expressed in the spleen, lymph nodes, and resting B cells, and to some extent in activated B cells, resting CD4+ cells and peripheral blood leukocytes. BAFFR is a type III transmembrane protein that binds with high specificity to BAFF (TNFSF13B). BAFFR/BAFF signaling plays a critical role in B cell survival and maturation. Recombinant Human BAFF Receptor is a 76 amino acid polypeptide (7.7 kDa) corresponding to the extracellular portion of the full BAFFR protein.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at –20 °C prior to reconstitution.
- Endotoxin content:>0.1 ng/μg
- Biological activity:Biological Activity tests can vary. Please refer to the Product CoA for information.
- Form:Lyophilized
- Gene ID:115650 Q96RJ3
- Reconstitution instructions:Reconstitution instructions can vary. Please refer to the product CoA for info.
- Protease-free:Y
- Protein synonyms:BAFFR|BLyS Receptor 3|CD268|TNFRSF13C
- Protein/peptide name:BAFF
- Purity:0.95
- Molecular weight:7.7 kDa
- Sequence:MRRGPRSLRG RDAPAPTPCV PAECFDLLVR HCVACGLLRT PRPKPAGASS PAPRTALQPQ ESVGAGAGEA ALPLPG
- Endotoxin level:Low
- Formulation:Formulation can vary. Please refer to product CoA for information.
- Shipping temperature:Ambient