Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Human Recombinant 4-1BB (from E. coli)

Supplier:   Peprotech
Human Recombinant 4-1BB (from <i>E. coli</i>)
Supplier:  Peprotech
Certificates

6 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

4-1BBL, a member of the TNF superfamily, is expressed in B cells, dendritic cells, activated T cells and macrophages. 4-1BBL binds to its receptor 4-1BB, and provides a co-stimulatory signal for T cell activation and expansion. The human 4-1BBL gene codes for a 254 amino acid type II transmembrane containing a 28 amino acid cytoplasmic domain, a 21 amino acid transmembrane domain, and a 205 amino acid extracellular domain. The soluble form of 4-1BBL contains the TNF-like portion of the extracellular domain of 4-1BBL. Recombinant Human 4-1BB Ligand is a soluble 19.5 kDa protein consisting of 185 amino acid residues.

 : For research use only. Not for use in diagnostic or therapeutic procedures.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 10774-198
  • 10774-200
  • 10774-202
  • 10774-204
  • 10774-206
  • 10774-208
  • Protein/peptide name:
  • 4-1BB
  • 4-1BB
  • 4-1BB
  • 4-1BB
  • 4-1BB
  • 4-1BB
  • Protein synonyms:
  • CD137L|TNFSF9
  • CD137L|TNFSF9
  • CD137L|TNFSF9
  • CD137L|TNFSF9
  • CD137L|TNFSF9
  • CD137L|TNFSF9
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein Function:
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Species:
  • Human
  • Human
  • Human
  • Human
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • 8744 P41273
  • 8744 P41273
  • 8744 P41273
  • 8744 P41273
  • 8744 P41273
  • 8744 P41273
  • Sequence:
  • MREGPELSPD DPAGLLDLRQ GMFAQLVAQN VLLIDGPLSW YSDPGLAGVS LTGGLSYKED TKELVVAKAG VYYVFFQLEL RRVVAGEGSG SVSLALHLQP LRSAAGAAAL ALTVDLPPAS SEARNSAFGF QGRLLHLSAG QRLGVHLHTE ARARHAWQLT QGATVLGLFR VTPEIPAGLP SPRSE
  • MREGPELSPD DPAGLLDLRQ GMFAQLVAQN VLLIDGPLSW YSDPGLAGVS LTGGLSYKED TKELVVAKAG VYYVFFQLEL RRVVAGEGSG SVSLALHLQP LRSAAGAAAL ALTVDLPPAS SEARNSAFGF QGRLLHLSAG QRLGVHLHTE ARARHAWQLT QGATVLGLFR VTPEIPAGLP SPRSE
  • MREGPELSPD DPAGLLDLRQ GMFAQLVAQN VLLIDGPLSW YSDPGLAGVS LTGGLSYKED TKELVVAKAG VYYVFFQLEL RRVVAGEGSG SVSLALHLQP LRSAAGAAAL ALTVDLPPAS SEARNSAFGF QGRLLHLSAG QRLGVHLHTE ARARHAWQLT QGATVLGLFR VTPEIPAGLP SPRSE
  • MREGPELSPD DPAGLLDLRQ GMFAQLVAQN VLLIDGPLSW YSDPGLAGVS LTGGLSYKED TKELVVAKAG VYYVFFQLEL RRVVAGEGSG SVSLALHLQP LRSAAGAAAL ALTVDLPPAS SEARNSAFGF QGRLLHLSAG QRLGVHLHTE ARARHAWQLT QGATVLGLFR VTPEIPAGLP SPRSE
  • MREGPELSPD DPAGLLDLRQ GMFAQLVAQN VLLIDGPLSW YSDPGLAGVS LTGGLSYKED TKELVVAKAG VYYVFFQLEL RRVVAGEGSG SVSLALHLQP LRSAAGAAAL ALTVDLPPAS SEARNSAFGF QGRLLHLSAG QRLGVHLHTE ARARHAWQLT QGATVLGLFR VTPEIPAGLP SPRSE
  • MREGPELSPD DPAGLLDLRQ GMFAQLVAQN VLLIDGPLSW YSDPGLAGVS LTGGLSYKED TKELVVAKAG VYYVFFQLEL RRVVAGEGSG SVSLALHLQP LRSAAGAAAL ALTVDLPPAS SEARNSAFGF QGRLLHLSAG QRLGVHLHTE ARARHAWQLT QGATVLGLFR VTPEIPAGLP SPRSE
  • Protease-free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Y
  • Endotoxin level:
  • Low
  • Low
  • Low
  • Low
  • Low
  • Low
  • Endotoxin content:
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • Biological activity:
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Purity:
  • 0.98
  • 0.98
  • 0.98
  • 0.98
  • 0.98
  • 0.98
  • Form:
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Formulation:
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Molecular weight:
  • 19.5 kDa
  • 19.5 kDa
  • 19.5 kDa
  • 19.5 kDa
  • 19.5 kDa
  • 19.5 kDa
  • Reconstitution instructions:
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Storage conditions:
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Shipping temperature:
  • Ambient
  • Ambient
  • Ambient
  • Ambient
  • Ambient
  • Ambient

Specifications

Product Family Options

Product Information

  • Size
    Origin
    Availability
    Price
  • 5 μg
    US
    Log in for availability
    Log in for availability
    Each (5µG)
    Each (5µG) - $248.04
    $248.04
    Each (5µG)
       $248.04
     

    Supplier Number: 310-11-5UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10774-198
    Protein/peptide name
    4-1BB
    Protein synonyms
    CD137L|TNFSF9
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    8744 P41273
    Sequence
    MREGPELSPD DPAGLLDLRQ GMFAQLVAQN VLLIDGPLSW YSDPGLAGVS LTGGLSYKED TKELVVAKAG VYYVFFQLEL RRVVAGEGSG SVSLALHLQP LRSAAGAAAL ALTVDLPPAS SEARNSAFGF QGRLLHLSAG QRLGVHLHTE ARARHAWQLT QGATVLGLFR VTPEIPAGLP SPRSE
    Protease-free
    Y
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    19.5 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient
  • 20 μg
    US
    Log in for availability
    Log in for availability
    Each (20µG)
    Each (20µG) - $558.42
    $558.42
    Each (20µG)
       $558.42
     

    Supplier Number: 310-11-20UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10774-200
    Protein/peptide name
    4-1BB
    Protein synonyms
    CD137L|TNFSF9
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    8744 P41273
    Sequence
    MREGPELSPD DPAGLLDLRQ GMFAQLVAQN VLLIDGPLSW YSDPGLAGVS LTGGLSYKED TKELVVAKAG VYYVFFQLEL RRVVAGEGSG SVSLALHLQP LRSAAGAAAL ALTVDLPPAS SEARNSAFGF QGRLLHLSAG QRLGVHLHTE ARARHAWQLT QGATVLGLFR VTPEIPAGLP SPRSE
    Protease-free
    Y
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    19.5 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient
  • 100 μg
    US
    Log in for availability
    Log in for availability
    Each (100µG)
    Each (100µG) - $1,788.84
    $1,788.84
    Each (100µG)
       $1,788.84
     

    Supplier Number: 310-11-100UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10774-202
    Protein/peptide name
    4-1BB
    Protein synonyms
    CD137L|TNFSF9
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    8744 P41273
    Sequence
    MREGPELSPD DPAGLLDLRQ GMFAQLVAQN VLLIDGPLSW YSDPGLAGVS LTGGLSYKED TKELVVAKAG VYYVFFQLEL RRVVAGEGSG SVSLALHLQP LRSAAGAAAL ALTVDLPPAS SEARNSAFGF QGRLLHLSAG QRLGVHLHTE ARARHAWQLT QGATVLGLFR VTPEIPAGLP SPRSE
    Protease-free
    Y
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    19.5 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient
  • 250 μg
    US
    Log in for availability
    Log in for availability
    Each (250µG)
    Each (250µG) - $2,787.04
    $2,787.04
    Each (250µG)
       $2,787.04
     

    Supplier Number: 310-11-250UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10774-204
    Protein/peptide name
    4-1BB
    Protein synonyms
    CD137L|TNFSF9
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    8744 P41273
    Sequence
    MREGPELSPD DPAGLLDLRQ GMFAQLVAQN VLLIDGPLSW YSDPGLAGVS LTGGLSYKED TKELVVAKAG VYYVFFQLEL RRVVAGEGSG SVSLALHLQP LRSAAGAAAL ALTVDLPPAS SEARNSAFGF QGRLLHLSAG QRLGVHLHTE ARARHAWQLT QGATVLGLFR VTPEIPAGLP SPRSE
    Protease-free
    Y
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    19.5 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient
  • 500 μg
    US
    Log in for availability
    Log in for availability
    Each (500µG)
    Each (500µG) - $4,777.24
    $4,777.24
    Each (500µG)
       $4,777.24
     

    Supplier Number: 310-11-500UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10774-206
    Protein/peptide name
    4-1BB
    Protein synonyms
    CD137L|TNFSF9
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    8744 P41273
    Sequence
    MREGPELSPD DPAGLLDLRQ GMFAQLVAQN VLLIDGPLSW YSDPGLAGVS LTGGLSYKED TKELVVAKAG VYYVFFQLEL RRVVAGEGSG SVSLALHLQP LRSAAGAAAL ALTVDLPPAS SEARNSAFGF QGRLLHLSAG QRLGVHLHTE ARARHAWQLT QGATVLGLFR VTPEIPAGLP SPRSE
    Protease-free
    Y
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    19.5 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient
  • 1 mg
    US
    Log in for availability
    Log in for availability
    Each (1mg)
    Each (1mg) - $7,969.24
    $7,969.24
    Each (1mg)
       $7,969.24
     

    Supplier Number: 310-11-1MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10774-208
    Protein/peptide name
    4-1BB
    Protein synonyms
    CD137L|TNFSF9
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    8744 P41273
    Sequence
    MREGPELSPD DPAGLLDLRQ GMFAQLVAQN VLLIDGPLSW YSDPGLAGVS LTGGLSYKED TKELVVAKAG VYYVFFQLEL RRVVAGEGSG SVSLALHLQP LRSAAGAAAL ALTVDLPPAS SEARNSAFGF QGRLLHLSAG QRLGVHLHTE ARARHAWQLT QGATVLGLFR VTPEIPAGLP SPRSE
    Protease-free
    Y
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    19.5 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient