Nesfatin-1 is a metabolic polypeptide encoded in the N-terminal region of the precursor protein, Nucleobindin2 (NUCB2). Originally identified as a hypothalamic neuropeptide, nesfatin-1 is also expressed in other areas of the brain, in pancreatic islets β-cells, gastric endocrine cells, and adipocytes. Nesfatin-1 suppresses food intake and can regulate energy metabolism in a Leptin independent manner. Recombinant Human Nesfatin-1 is a 9.7 kDa protein containing 82 amino acid residues.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at –20 °C prior to reconstitution.
- Endotoxin content:>0.1 ng/μg
- Biological activity:Biological Activity tests can vary. Please refer to the Product CoA for information.
- Form:Lyophilized
- Gene ID:4925 P80303
- Reconstitution instructions:Reconstitution instructions can vary. Please refer to the product CoA for info.
- Protease-free:Y
- Protein/peptide name:Nesfatin-1
- Purity:0.98
- Molecular weight:9.7 kDa
- Sequence:VPIDIDKTKV QNIHPVESAK IEPPDTGLYY DEYLKQVIDV LETDKHFREK LQKADIEEIK SGRLSKELDL VSHHVRTKLD EL
- Endotoxin level:Low
- Formulation:Formulation can vary. Please refer to product CoA for information.
- Shipping temperature:Ambient