Human Recombinant TAFA-2 (from E. coli)
Supplier: Peprotech
Certificates
About this item
TAFA proteins are a newly discovered family of proteins, which are distantly related to MIP-1α, a member of the CC-chemokine family. TAFA mRNAs are highly expressed in specific brain regions. The biological function of TAFA-2 is still unknown. Recombinant Human TAFA-2 is an 11.2 kDa protein consisting of 101 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at –20 °C prior to reconstitution.
- Endotoxin content:>0.1 ng/μg
- Biological activity:Biological Activity tests can vary. Please refer to the Product CoA for information.
- Form:Lyophilized
- Gene ID:338811 Q8N3H0
- Reconstitution instructions:Reconstitution instructions can vary. Please refer to the product CoA for info.
- Protease-free:Y
- Protein synonyms:FAM19A2|Chemokine-like protein TAFA-2
- Protein/peptide name:TAFA-2
- Purity:0.98
- Molecular weight:11.2 kDa
- Sequence:ANHHKAHHVK TGTCEVVALH RCCNKNKIEE RSQTVKCSCF PGQVAGTTRA APSCVDASIV EQKWWCHMQP CLEGEECKVL PDRKGWSCSS GNKVKTTRVT H
- Endotoxin level:Low
- Formulation:Formulation can vary. Please refer to product CoA for information.
- Shipping temperature:Ambient
Specifications
Frequently Bought Together