Human Recombinant MIP-3beta (from E. coli)
Supplier: Peprotech
Certificates
About this item
MIP-3β is a CC chemokine, expressed in the thymus, lymph nodes, and in activated bone marrow stromal cells that signals through the CCR7 receptor. MIP-3β is a chemoattractant for T and B lymphocytes, and myeloid progenitor cells. Human MIP-3β is active on murine cells. Recombinant Human MIP-3β (CCL19) is an 8.8 kDa protein containing 77 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at –20 °C prior to reconstitution.
- Endotoxin content:>0.1 ng/μg
- Biological activity:Biological Activity tests can vary. Please refer to the Product CoA for information.
- Form:Lyophilized
- Gene ID:6363 Q99731
- Reconstitution instructions:Reconstitution instructions can vary. Please refer to the product CoA for info.
- Protease-free:Y
- Protein synonyms:SCYA19|ELC|Macrophage Inflammatory Protein-3β|Exodus-3
- Protein/peptide name:MIP-3beta
- Purity:0.98
- Molecular weight:8.8 kDa
- Sequence:GTNDAEDCCL SVTQKPIPGY IVRNFHYLLI KDGCRVPAVV FTTLRGRQLC APPDQPWVER IIQRLQRTSA KMKRRSS
- Endotoxin level:Low
- Formulation:Formulation can vary. Please refer to product CoA for information.
- Shipping temperature:Ambient
Specifications
Frequently Bought Together