Human Recombinant PF-4 (from E. coli)
PF-4 is a CXC chemokine that is expressed in megakaryocytes and stored in the α-granules of platelets. PF-4 is chemotactic towards neutrophils and monocytes, and has been shown to inhibit angiogenesis. Recombinant Human PF-4 (CXCL4) is a 7.8 kDa protein containing 70 amino acid residues, including the four highly conserved residues present in CXC chemokines.
: For research use only. Not for use in diagnostic or therapeutic procedures.
PF-4 is a CXC chemokine that is expressed in megakaryocytes and stored in the α-granules of platelets. PF-4 is chemotactic towards neutrophils and monocytes, and has been shown to inhibit angiogenesis. Recombinant Human PF-4 (CXCL4) is a 7.8 kDa protein containing 70 amino acid residues, including the four highly conserved residues present in CXC chemokines.
: For research use only. Not for use in diagnostic or therapeutic procedures.
- Catalog No:
- 10773-570
- 10773-572
- 10773-574
- 10773-576
- 10773-578
- 10773-580
- 10773-582
- Protein/peptide name:
- PF-4
- PF-4
- PF-4
- PF-4
- PF-4
- PF-4
- PF-4
- Protein synonyms:
- Oncostatin A|Iroplact|Platelet Factor-4
- Oncostatin A|Iroplact|Platelet Factor-4
- Oncostatin A|Iroplact|Platelet Factor-4
- Oncostatin A|Iroplact|Platelet Factor-4
- Oncostatin A|Iroplact|Platelet Factor-4
- Oncostatin A|Iroplact|Platelet Factor-4
- Oncostatin A|Iroplact|Platelet Factor-4
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein Function:
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Species:
- Human
- Human
- Human
- Human
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- 5196 P02776
- 5196 P02776
- 5196 P02776
- 5196 P02776
- 5196 P02776
- 5196 P02776
- 5196 P02776
- Sequence:
- EAEEDGDLQC LCVKTTSQVR PRHITSLEVI KAGPHCPTAQ LIATLKNGRK ICLDLQAPLY KKIIKKLLES
- EAEEDGDLQC LCVKTTSQVR PRHITSLEVI KAGPHCPTAQ LIATLKNGRK ICLDLQAPLY KKIIKKLLES
- EAEEDGDLQC LCVKTTSQVR PRHITSLEVI KAGPHCPTAQ LIATLKNGRK ICLDLQAPLY KKIIKKLLES
- EAEEDGDLQC LCVKTTSQVR PRHITSLEVI KAGPHCPTAQ LIATLKNGRK ICLDLQAPLY KKIIKKLLES
- EAEEDGDLQC LCVKTTSQVR PRHITSLEVI KAGPHCPTAQ LIATLKNGRK ICLDLQAPLY KKIIKKLLES
- EAEEDGDLQC LCVKTTSQVR PRHITSLEVI KAGPHCPTAQ LIATLKNGRK ICLDLQAPLY KKIIKKLLES
- EAEEDGDLQC LCVKTTSQVR PRHITSLEVI KAGPHCPTAQ LIATLKNGRK ICLDLQAPLY KKIIKKLLES
- Protease-free:
- Y
- Y
- Y
- Y
- Y
- Y
- Y
- Endotoxin level:
- Low
- Low
- Low
- Low
- Low
- Low
- Low
- Endotoxin content:
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- Biological activity:
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Purity:
- 0.98
- 0.98
- 0.98
- 0.98
- 0.98
- 0.98
- 0.98
- Form:
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Formulation:
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Molecular weight:
- 7.8 kDa
- 7.8 kDa
- 7.8 kDa
- 7.8 kDa
- 7.8 kDa
- 7.8 kDa
- 7.8 kDa
- Reconstitution instructions:
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Storage conditions:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping temperature:
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
Product Family Options
- SizeOrigin
- : 300-16-5UG
Specifications
Cat. No.10773-570Protein/peptide namePF-4Protein synonymsOncostatin A|Iroplact|Platelet Factor-4Protein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID5196 P02776SequenceEAEEDGDLQC LCVKTTSQVR PRHITSLEVI KAGPHCPTAQ LIATLKNGRK ICLDLQAPLY KKIIKKLLESProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight7.8 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - : 300-16-20UG
Specifications
Cat. No.10773-572Protein/peptide namePF-4Protein synonymsOncostatin A|Iroplact|Platelet Factor-4Protein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID5196 P02776SequenceEAEEDGDLQC LCVKTTSQVR PRHITSLEVI KAGPHCPTAQ LIATLKNGRK ICLDLQAPLY KKIIKKLLESProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight7.8 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - : 300-16-50UG
Specifications
Cat. No.10773-574Protein/peptide namePF-4Protein synonymsOncostatin A|Iroplact|Platelet Factor-4Protein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID5196 P02776SequenceEAEEDGDLQC LCVKTTSQVR PRHITSLEVI KAGPHCPTAQ LIATLKNGRK ICLDLQAPLY KKIIKKLLESProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight7.8 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - : 300-16-100UG
Specifications
Cat. No.10773-576Protein/peptide namePF-4Protein synonymsOncostatin A|Iroplact|Platelet Factor-4Protein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID5196 P02776SequenceEAEEDGDLQC LCVKTTSQVR PRHITSLEVI KAGPHCPTAQ LIATLKNGRK ICLDLQAPLY KKIIKKLLESProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight7.8 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - : 300-16-250UG
Specifications
Cat. No.10773-578Protein/peptide namePF-4Protein synonymsOncostatin A|Iroplact|Platelet Factor-4Protein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID5196 P02776SequenceEAEEDGDLQC LCVKTTSQVR PRHITSLEVI KAGPHCPTAQ LIATLKNGRK ICLDLQAPLY KKIIKKLLESProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight7.8 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - : 300-16-500UG
Specifications
Cat. No.10773-580Protein/peptide namePF-4Protein synonymsOncostatin A|Iroplact|Platelet Factor-4Protein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID5196 P02776SequenceEAEEDGDLQC LCVKTTSQVR PRHITSLEVI KAGPHCPTAQ LIATLKNGRK ICLDLQAPLY KKIIKKLLESProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight7.8 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - : 300-16-1MG
Specifications
Cat. No.10773-582Protein/peptide namePF-4Protein synonymsOncostatin A|Iroplact|Platelet Factor-4Protein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesHumanSourceE. coliConjugationUnconjugatedGene ID5196 P02776SequenceEAEEDGDLQC LCVKTTSQVR PRHITSLEVI KAGPHCPTAQ LIATLKNGRK ICLDLQAPLY KKIIKKLLESProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight7.8 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient



