Mouse Recombinant SDF-1alpha (from E. coli)
SDF-1α and β are stromal-derived, CXC chemokines that signal through the CXCR4 receptor. SDF-1α and β chemoattract B and T cells, and have been shown to induce migration of CD34+ stem cells. Additionally, the SDF-1 proteins exert HIV-suppressive activity in cells expressing the CXCR4 receptor. Human and murine SDF-1 proteins act across species. SDF-1α and β contain the four highly conserved cysteine residues present in CXC chemokines. The mature SDF-1α protein is the result of alternative splicing of the SDF-1 gene and contains 68 amino acid residues. Recombinant Murine SDF-1α (CXCL12) is a 7.9 kDa protein containing 68 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
SDF-1α and β are stromal-derived, CXC chemokines that signal through the CXCR4 receptor. SDF-1α and β chemoattract B and T cells, and have been shown to induce migration of CD34+ stem cells. Additionally, the SDF-1 proteins exert HIV-suppressive activity in cells expressing the CXCR4 receptor. Human and murine SDF-1 proteins act across species. SDF-1α and β contain the four highly conserved cysteine residues present in CXC chemokines. The mature SDF-1α protein is the result of alternative splicing of the SDF-1 gene and contains 68 amino acid residues. Recombinant Murine SDF-1α (CXCL12) is a 7.9 kDa protein containing 68 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
- Catalog No:
- 10773-274
- 10773-276
- 10773-278
- Protein/peptide name:
- SDF-1alpha
- SDF-1alpha
- SDF-1alpha
- Protein synonyms:
- PBSF|Stromal-Cell Derived Factor-1
- PBSF|Stromal-Cell Derived Factor-1
- PBSF|Stromal-Cell Derived Factor-1
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Protein Function:
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Cytokines and Growth Factors
- Species:
- Mouse
- Mouse
- Mouse
- Source:
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- 20315 P40224
- 20315 P40224
- 20315 P40224
- Sequence:
- KPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNK
- KPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNK
- KPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNK
- Protease-free:
- Y
- Y
- Y
- Endotoxin level:
- Low
- Low
- Low
- Endotoxin content:
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- Biological activity:
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Purity:
- 0.98
- 0.98
- 0.98
- Form:
- Lyophilized
- Lyophilized
- Lyophilized
- Formulation:
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Molecular weight:
- 7.9 kDa
- 7.9 kDa
- 7.9 kDa
- Reconstitution instructions:
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Storage conditions:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping temperature:
- Ambient
- Ambient
- Ambient
Product Family Options
- SizeOrigin
- : 250-20A-250UG
Specifications
Cat. No.10773-274Protein/peptide nameSDF-1alphaProtein synonymsPBSF|Stromal-Cell Derived Factor-1Protein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID20315 P40224SequenceKPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight7.9 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - : 250-20A-500UG
Specifications
Cat. No.10773-276Protein/peptide nameSDF-1alphaProtein synonymsPBSF|Stromal-Cell Derived Factor-1Protein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID20315 P40224SequenceKPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight7.9 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - : 250-20A-1MG
Specifications
Cat. No.10773-278Protein/peptide nameSDF-1alphaProtein synonymsPBSF|Stromal-Cell Derived Factor-1Protein/peptide typeRecombinantProtein FunctionCytokines and Growth FactorsSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID20315 P40224SequenceKPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight7.9 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient



