Order Entry
Canada
Orders LinkContactUsLinkComponent
Mouse Recombinant LIX (from <i>E. coli</i>)

Mouse Recombinant LIX (from E. coli)

Supplier:   Peprotech
Mouse Recombinant LIX (from <i>E. coli</i>)
Supplier:  Peprotech
Certificates

6 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

CXCL6, also known as LIX in mice, is a CXC chemokine that signals through the CXCR2 receptor. It is expressed in monocytes, platelets, endothelial cells, and mast cells. LIX is a chemoattractant for neutrophils. The two naturally occurring variants of LIX, LIX 1-78 (GCP-2) and LIX 9-78 (GCP-2), contain 78 and 70 amino acid residues, respectively. LIX contains the four conserved cysteine residues present in CXC chemokines, and also contains the ‘ELR’ motif common to CXC chemokines that binds to the CXCR1 and CXCR2 receptors. Recombinant Murine LIX (CXCL6) is a 9.8 kDa protein containing 92 amino acid residues.

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 10773-254
  • 10773-256
  • 10773-258
  • 10773-260
  • 10773-262
  • 10773-264
  • Protein/peptide name:
  • LIX
  • LIX
  • LIX
  • LIX
  • LIX
  • LIX
  • Protein synonyms:
  • Granulocyte Chemotactic Protein-2
  • Granulocyte Chemotactic Protein-2
  • Granulocyte Chemotactic Protein-2
  • Granulocyte Chemotactic Protein-2
  • Granulocyte Chemotactic Protein-2
  • Granulocyte Chemotactic Protein-2
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein Function:
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Cytokines and Growth Factors
  • Species:
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • 20311 P50228
  • 20311 P50228
  • 20311 P50228
  • 20311 P50228
  • 20311 P50228
  • 20311 P50228
  • Sequence:
  • APSSVIAATE LRCVCLTVTP KINPKLIANL EVIPAGPQCP TVEVIAKLKN QKEVCLDPEA PVIKKIIQKI LGSDKKKAKR NALAVERTAS VQ
  • APSSVIAATE LRCVCLTVTP KINPKLIANL EVIPAGPQCP TVEVIAKLKN QKEVCLDPEA PVIKKIIQKI LGSDKKKAKR NALAVERTAS VQ
  • APSSVIAATE LRCVCLTVTP KINPKLIANL EVIPAGPQCP TVEVIAKLKN QKEVCLDPEA PVIKKIIQKI LGSDKKKAKR NALAVERTAS VQ
  • APSSVIAATE LRCVCLTVTP KINPKLIANL EVIPAGPQCP TVEVIAKLKN QKEVCLDPEA PVIKKIIQKI LGSDKKKAKR NALAVERTAS VQ
  • APSSVIAATE LRCVCLTVTP KINPKLIANL EVIPAGPQCP TVEVIAKLKN QKEVCLDPEA PVIKKIIQKI LGSDKKKAKR NALAVERTAS VQ
  • APSSVIAATE LRCVCLTVTP KINPKLIANL EVIPAGPQCP TVEVIAKLKN QKEVCLDPEA PVIKKIIQKI LGSDKKKAKR NALAVERTAS VQ
  • Protease-free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Y
  • Endotoxin level:
  • Low
  • Low
  • Low
  • Low
  • Low
  • Low
  • Endotoxin content:
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • >0.1 ng/μg
  • Biological activity:
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Purity:
  • 0.98
  • 0.98
  • 0.98
  • 0.98
  • 0.98
  • 0.98
  • Form:
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Formulation:
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Formulation can vary. Please refer to product CoA for information.
  • Molecular weight:
  • 9.8 kDa
  • 9.8 kDa
  • 9.8 kDa
  • 9.8 kDa
  • 9.8 kDa
  • 9.8 kDa
  • Reconstitution instructions:
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Storage conditions:
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Shipping temperature:
  • Ambient
  • Ambient
  • Ambient
  • Ambient
  • Ambient
  • Ambient

Specifications

Product Family Options

Product Information

  • Size
    Origin
    Availability
    Price
  • 5 μg
    US
    Log in for availability
    Log in for availability
    Each (5µG)
    Each (5µG) - $241.16
    $241.16
    Each (5µG)
       $241.16
    Minimum order quantity is 1

    Supplier Number: 250-17-5UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10773-254
    Protein/peptide name
    LIX
    Protein synonyms
    Granulocyte Chemotactic Protein-2
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    20311 P50228
    Sequence
    APSSVIAATE LRCVCLTVTP KINPKLIANL EVIPAGPQCP TVEVIAKLKN QKEVCLDPEA PVIKKIIQKI LGSDKKKAKR NALAVERTAS VQ
    Protease-free
    Y
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    9.8 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient
  • 20 μg
    US
    Log in for availability
    Log in for availability
    Each (20µG)
    Each (20µG) - $582.62
    $582.62
    Each (20µG)
       $582.62
    Minimum order quantity is 1

    Supplier Number: 250-17-20UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10773-256
    Protein/peptide name
    LIX
    Protein synonyms
    Granulocyte Chemotactic Protein-2
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    20311 P50228
    Sequence
    APSSVIAATE LRCVCLTVTP KINPKLIANL EVIPAGPQCP TVEVIAKLKN QKEVCLDPEA PVIKKIIQKI LGSDKKKAKR NALAVERTAS VQ
    Protease-free
    Y
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    9.8 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient
  • 100 μg
    US
    Log in for availability
    Log in for availability
    Each (100µG)
    Each (100µG) - $1,716.42
    $1,716.42
    Each (100µG)
       $1,716.42
    Minimum order quantity is 1

    Supplier Number: 250-17-100UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10773-258
    Protein/peptide name
    LIX
    Protein synonyms
    Granulocyte Chemotactic Protein-2
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    20311 P50228
    Sequence
    APSSVIAATE LRCVCLTVTP KINPKLIANL EVIPAGPQCP TVEVIAKLKN QKEVCLDPEA PVIKKIIQKI LGSDKKKAKR NALAVERTAS VQ
    Protease-free
    Y
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    9.8 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient
  • 250 μg
    US
    Log in for availability
    Log in for availability
    Each (250µG)
    Each (250µG) - $2,787.04
    $2,787.04
    Each (250µG)
       $2,787.04
    Minimum order quantity is 1

    Supplier Number: 250-17-250UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10773-260
    Protein/peptide name
    LIX
    Protein synonyms
    Granulocyte Chemotactic Protein-2
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    20311 P50228
    Sequence
    APSSVIAATE LRCVCLTVTP KINPKLIANL EVIPAGPQCP TVEVIAKLKN QKEVCLDPEA PVIKKIIQKI LGSDKKKAKR NALAVERTAS VQ
    Protease-free
    Y
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    9.8 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient
  • 500 μg
    US
    Log in for availability
    Log in for availability
    Each (500µG)
    Each (500µG) - $4,788.08
    $4,788.08
    Each (500µG)
       $4,788.08
    Minimum order quantity is 1

    Supplier Number: 250-17-500UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10773-262
    Protein/peptide name
    LIX
    Protein synonyms
    Granulocyte Chemotactic Protein-2
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    20311 P50228
    Sequence
    APSSVIAATE LRCVCLTVTP KINPKLIANL EVIPAGPQCP TVEVIAKLKN QKEVCLDPEA PVIKKIIQKI LGSDKKKAKR NALAVERTAS VQ
    Protease-free
    Y
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    9.8 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient
  • 1 mg
    US
    Log in for availability
    Log in for availability
    Each (1mg)
    Each (1mg) - $7,969.24
    $7,969.24
    Each (1mg)
       $7,969.24
    Minimum order quantity is 1

    Supplier Number: 250-17-1MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10773-264
    Protein/peptide name
    LIX
    Protein synonyms
    Granulocyte Chemotactic Protein-2
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokines and Growth Factors
    Species
    Mouse
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    20311 P50228
    Sequence
    APSSVIAATE LRCVCLTVTP KINPKLIANL EVIPAGPQCP TVEVIAKLKN QKEVCLDPEA PVIKKIIQKI LGSDKKKAKR NALAVERTAS VQ
    Protease-free
    Y
    Endotoxin level
    Low
    Endotoxin content
    >0.1 ng/μg
    Biological activity
    Biological Activity tests can vary. Please refer to the Product CoA for information.
    Purity
    0.98
    Form
    Lyophilized
    Formulation
    Formulation can vary. Please refer to product CoA for information.
    Molecular weight
    9.8 kDa
    Reconstitution instructions
    Reconstitution instructions can vary. Please refer to the product CoA for info.
    Storage conditions
    Store at –20 °C prior to reconstitution.
    Shipping temperature
    Ambient
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.