Order Entry
Canada
ContactUsLinkComponent
Human Recombinant BMP-6 (from HEK293 cells)
Human Recombinant BMP-6 (from HEK293 cells)
Supplier:  Peprotech
Certificates
Human Recombinant BMP-6 (from HEK293 cells)
Supplier:  Peprotech

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C prior to reconstitution.
  • Endotoxin content:
    >0.1 ng/μg
  • Biological activity:
    Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Form:
    Lyophilized
  • Gene ID:
    654 P22004
  • Reconstitution instructions:
    Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Protease-free:
    Y
  • Protein synonyms:
    VGR|VG-1-related protein|Bone Morphogenetic Protein-6
  • Protein/peptide name:
    BMP-6
  • Purity:
    0.95
  • Molecular weight:
    26.2 kDa
  • Sequence:
    VSSASDYNSS ELKTACRKHE LYVSFQDLGW QDWIIAPKGY AANYCDGECS FPLNAHMNAT NHAIVQTLVH LMNPEYVPKP CCAPTKLNAI SVLYFDDNSN VILKKYRNMV VRACGCH
  • Endotoxin level:
    Low
  • Formulation:
    Formulation can vary. Please refer to product CoA for information.
  • Shipping temperature:
    Ambient

Specifications

Frequently Bought Together