Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Rat Recombinant IL2 (from E. coli)

Supplier:   Peprotech
Rat Recombinant IL2 (from <i>E. coli</i>)
Supplier:  Peprotech
Certificates

6 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

IL-2 is a powerful immunoregulatory lymphokine produced by T-cells in response to antigenic or mitogenic stimulation.  IL-2/IL-2R signaling is required for T-cell proliferation and other fundamental functions that are essential for the immune response.  IL-2 stimulates growth and differentiation of B-cells, NK cells, lymphokine-activated killer cells, monocytes, macrophages and oligodendrocytes.  Recombinant Rat IL-2 is a 15.3 kDa protein containing 134 amino acid residues. Manufactured using all Animal-Free reagents.

  • Carrier free
  • Animal-free

MAPTSSPAKE TQQHLEQLLL DLQVLLRGID NYKNLKLPMM LTFKFYLPKQ ATELKHLQCL ENELGALQRV LDLTQSKSFH LEDAGNFISN IRVTVVKLKG SENKFECQFD DEPATVVEFL RRWIAIAQSI ISTM

Verified by N-terminal and Mass Spectrometry analyses (when applicable).

 : For research use only. Not for use in diagnostic or therapeutic procedures.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 10771-786
  • 10771-788
  • 10771-790
  • 10771-792
  • 10771-794
  • 10771-796
  • Protein/peptide name:
  • IL2
  • IL2
  • IL2
  • IL2
  • IL2
  • IL2
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Species:
  • Rat
  • Rat
  • Rat
  • Rat
  • Rat
  • Rat
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Animal-free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Y
  • Endotoxin content:
  • <0.1 ng/μg of protein (<1 EU/μg).
  • <0.1 ng/μg of protein (<1 EU/μg).
  • <0.1 ng/μg of protein (<1 EU/μg).
  • <0.1 ng/μg of protein (<1 EU/μg).
  • <0.1 ng/μg of protein (<1 EU/μg).
  • <0.1 ng/μg of protein (<1 EU/μg).
  • Biological activity:
  • The ED50 as determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is ≤ 0.4 ng/ml, corresponding to a specific activity of ≥ 2.5 x 106 units/mg.
  • The ED50 as determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is ≤ 0.4 ng/ml, corresponding to a specific activity of ≥ 2.5 x 106 units/mg.
  • The ED50 as determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is ≤ 0.4 ng/ml, corresponding to a specific activity of ≥ 2.5 x 106 units/mg.
  • The ED50 as determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is ≤ 0.4 ng/ml, corresponding to a specific activity of ≥ 2.5 x 106 units/mg.
  • The ED50 as determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is ≤ 0.4 ng/ml, corresponding to a specific activity of ≥ 2.5 x 106 units/mg.
  • The ED50 as determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is ≤ 0.4 ng/ml, corresponding to a specific activity of ≥ 2.5 x 106 units/mg.
  • Purity:
  • Greater than 98% by SDS-PAGE gel and HPLC analyses.
  • Greater than 98% by SDS-PAGE gel and HPLC analyses.
  • Greater than 98% by SDS-PAGE gel and HPLC analyses.
  • Greater than 98% by SDS-PAGE gel and HPLC analyses.
  • Greater than 98% by SDS-PAGE gel and HPLC analyses.
  • Greater than 98% by SDS-PAGE gel and HPLC analyses.
  • Formulation:
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Storage conditions:
  • −20 °C
  • −20 °C
  • −20 °C
  • −20 °C
  • −20 °C
  • −20 °C
  • Shipping temperature:
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT

Specifications

Product Family Options

Product Information

  • Size
    Origin
    Availability
    Price
  • 5 µg
    US
    Log in for availability
    Log in for availability
    Each (5µG)
    Each (5µG) - $315.11
    $315.11
    Each (5µG)
       $315.11
     

    Supplier Number: AF-400-02-5UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10771-786
    Protein/peptide name
    IL2
    Protein/peptide type
    Recombinant
    Species
    Rat
    Source
    E. coli
    Animal-free
    Y
    Endotoxin content
    <0.1 ng/μg of protein (<1 EU/μg).
    Biological activity
    The ED50 as determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is ≤ 0.4 ng/ml, corresponding to a specific activity of ≥ 2.5 x 106 units/mg.
    Purity
    Greater than 98% by SDS-PAGE gel and HPLC analyses.
    Formulation
    Lyophilized
    Storage conditions
    −20 °C
    Shipping temperature
    RT
  • 20 µg
    US
    Log in for availability
    Log in for availability
    Each (20µG)
    Each (20µG) - $709.04
    $709.04
    Each (20µG)
       $709.04
     

    Supplier Number: AF-400-02-20UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10771-788
    Protein/peptide name
    IL2
    Protein/peptide type
    Recombinant
    Species
    Rat
    Source
    E. coli
    Animal-free
    Y
    Endotoxin content
    <0.1 ng/μg of protein (<1 EU/μg).
    Biological activity
    The ED50 as determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is ≤ 0.4 ng/ml, corresponding to a specific activity of ≥ 2.5 x 106 units/mg.
    Purity
    Greater than 98% by SDS-PAGE gel and HPLC analyses.
    Formulation
    Lyophilized
    Storage conditions
    −20 °C
    Shipping temperature
    RT
  • 100 µg
    US
    Log in for availability
    Log in for availability
    Each (100µG)
    Each (100µG) - $2,355.00
    $2,355.00
    Each (100µG)
       $2,355.00
     

    Supplier Number: AF-400-02-100UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10771-790
    Protein/peptide name
    IL2
    Protein/peptide type
    Recombinant
    Species
    Rat
    Source
    E. coli
    Animal-free
    Y
    Endotoxin content
    <0.1 ng/μg of protein (<1 EU/μg).
    Biological activity
    The ED50 as determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is ≤ 0.4 ng/ml, corresponding to a specific activity of ≥ 2.5 x 106 units/mg.
    Purity
    Greater than 98% by SDS-PAGE gel and HPLC analyses.
    Formulation
    Lyophilized
    Storage conditions
    −20 °C
    Shipping temperature
    RT
  • 250 µg
    US
    Log in for availability
    Log in for availability
    Each (250µG)
    Each (250µG) - $3,016.85
    $3,016.85
    Each (250µG)
       $3,016.85
     

    Supplier Number: AF-400-02-250UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10771-792
    Protein/peptide name
    IL2
    Protein/peptide type
    Recombinant
    Species
    Rat
    Source
    E. coli
    Animal-free
    Y
    Endotoxin content
    <0.1 ng/μg of protein (<1 EU/μg).
    Biological activity
    The ED50 as determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is ≤ 0.4 ng/ml, corresponding to a specific activity of ≥ 2.5 x 106 units/mg.
    Purity
    Greater than 98% by SDS-PAGE gel and HPLC analyses.
    Formulation
    Lyophilized
    Storage conditions
    −20 °C
    Shipping temperature
    RT
  • 500 µg
    US
    Log in for availability
    Log in for availability
    Each (500µG)
    Each (500µG) - $5,174.07
    $5,174.07
    Each (500µG)
       $5,174.07
     

    Supplier Number: AF-400-02-500UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10771-794
    Protein/peptide name
    IL2
    Protein/peptide type
    Recombinant
    Species
    Rat
    Source
    E. coli
    Animal-free
    Y
    Endotoxin content
    <0.1 ng/μg of protein (<1 EU/μg).
    Biological activity
    The ED50 as determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is ≤ 0.4 ng/ml, corresponding to a specific activity of ≥ 2.5 x 106 units/mg.
    Purity
    Greater than 98% by SDS-PAGE gel and HPLC analyses.
    Formulation
    Lyophilized
    Storage conditions
    −20 °C
    Shipping temperature
    RT
  • 1 mg
    US
    Log in for availability
    Log in for availability
    Each (1mg)
    Each (1mg) - $8,624.51
    $8,624.51
    Each (1mg)
       $8,624.51
     

    Supplier Number: AF-400-02-1MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10771-796
    Protein/peptide name
    IL2
    Protein/peptide type
    Recombinant
    Species
    Rat
    Source
    E. coli
    Animal-free
    Y
    Endotoxin content
    <0.1 ng/μg of protein (<1 EU/μg).
    Biological activity
    The ED50 as determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is ≤ 0.4 ng/ml, corresponding to a specific activity of ≥ 2.5 x 106 units/mg.
    Purity
    Greater than 98% by SDS-PAGE gel and HPLC analyses.
    Formulation
    Lyophilized
    Storage conditions
    −20 °C
    Shipping temperature
    RT