About this item
Proteases (also called Proteolytic Enzymes, Peptidases, or Proteinases) are enzymes that hydrolyze the amide bonds within proteins or peptides. Most proteases act in a specific manner, hydrolyzing bonds at, or adjacent to specific residues, or a specific sequence of residues contained within the substrate protein or peptide. Proteases play an important role in most diseases and biological processes, including prenatal and postnatal development, reproduction, signal transduction, the immune response, various autoimmune and degenerative diseases, and cancer. They are also an important research tool, frequently used in the analysis and production of proteins. Recombinant Aeromonas Aminopeptidase is a 31.4 kDa protein containing 291 amino acid residues.
- Origin: USA
- Accession number: Q01693
Specifications
- Conjugation:Unconjugated
- Enzyme type:Recombinant
- Source:E. coli
- Species:Aeromonas
- Storage conditions:Store at –20 °C prior to reconstitution.
- Endotoxin content:>0.1 ng/μg
- Form:Lyophilized
- Shipping conditions:Ambient
- Reconstitution instructions:Reconstitution instructions can vary. Please refer to the product CoA for info.
- Enzyme name:Areomonas Aminopeptidase
- Enzyme activity:Biological Activity tests can vary. Please refer to the Product CoA for information.
- Purity:98%
- Molecular weight:31.4 kDa
- Sequence:MPPITQQATV TAWLPQVDAS QITGTISSLE SFTNRFYTTT SGAQASDWIA SEWQALSASL PNASVKQVSH SGYNQKSVVM TITGSEAPDE WIVIGGHLDS TIGSHTNEQS VAPGADDDAS GIAAVTEVIR VLSENNFQPK RSIAFMAYAA EEVGLRGSQD LANQYKSEGK NVVSALQLDM TNYKGSAQDV VFITDYTDSN FTQYLTQLMD EYLPSLTYGF DTCGYACSDH ASWHNAGYPA AMPFESKFND YNPRIHTTQD TLANSDPTGS HAKKFTQLGL AYAIEMGSAT G
- Endotoxin level:Low
- Formulation:Formulation can vary. Please refer to product CoA for information.