To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen name:SWI/SNF related Matrix Associated Actin Dependent Regulator of Chromatin Subfamily A member 1
- Antigen symbol:SMARCA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:6594
- Antigen synonyms:SWI2|SNF2LT|SWI|SNF2L|SNF2L1|ISWI|NURF140|SNF2LB
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10428-556
- Supplier No.:BS-6015R
SNF2L, also known as SMARCA1 (SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 1), SWI or ISWI, is a 1,054 amino acid protein that localizes to the nucleus and contains one helicase C-terminal domain, one helicase ATP-binding domain and two SANT domains. Expressed as multiple alternatively spliced isoforms, SNF2L exists as a component of the nucleosome-remodeling factor (NURF) complex where it helps to facilitate the ATP-dependent perturbation of chromatin structure and may also be involved in brain development and neurite outgrowth. The gene encoding SNF2L maps to human chromosome X, which contains nearly 153 million base pairs and houses over 1,000 genes.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: SMARCA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat