Order Entry
Canada
ContactUsLinkComponent
Anti-DCUN1D4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-DCUN1D4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
  10480-274
 :  Bioss
Anti-DCUN1D4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
  10480-274
 :  Bioss
 :  BS-8251R-HRP

 

  • Antibody type:
    Primary
  • Antigen name:
    DCN1, defective in cullin neddylation 1, Domain Containing 4
  • Antigen symbol:
    DCUN1D4
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µL
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    23142
  • Antigen synonyms:
    KIAA0276|DCUN1 domain containing protein 4|DCNL4_HUMAN.|DCN1 like protein 4|DCUN1D 4
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Cat. no.:
    10480-274
  • Supplier No.:
    BS-8251R-HRP

 

 

The DCN1-like protein family is comprised of Dcun1D1, Dcun1D2, Dcun1D3, Dcun1D4 and Dcun1D5. The founding member, Dcun1D1, is involved in the malignant transformation of squamous cell lineage.Dcun1D4, (defective in cullin neddylation protein 1-like protein 4 or DCN1-like protein 4), also designated KIAA0276, exists as 2 isoforms as a result of alternative splicing and contains one DCUN1 domain. The gene encoding Dcun1D4 maps to chromosome 4, which houses nearly 6% of the human genome and has the largest gene deserts (regions of the genome with no protein encoding genes) of all of the human chromosomes. Defects in some of the genes located on chromosome 4 are associated with Huntington's disease, Ellis-van Creveld syndrome, methylmalonic acidemia and polycystic kidney disease.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: DCUN1D4
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat