Order Entry
Canada
ContactUsLinkComponent
Anti-LAP3 Rabbit Polyclonal Antibody (Cy7®)
Anti-LAP3 Rabbit Polyclonal Antibody (Cy7®)
Catalog # 10467-456
Supplier:  Bioss
Anti-LAP3 Rabbit Polyclonal Antibody (Cy7®)
Catalog # 10467-456
Supplier:  Bioss
Supplier Number:  BS-7687R-CY7

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Leucine aminopeptidase 3
  • Antigen symbol:
    LAP3
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 µL
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    51056
  • Antigen synonyms:
    Prolyl aminopeptidase|Leucine aminopeptidase|Peptidase S|Proline aminopeptidase|Cytosol aminopeptidase|Leucyl aminopeptidase|LAP|Leucine aminopeptidase 3|LAP3
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Cat. no.:
    10467-456
  • Supplier No.:
    BS-7687R-CY7

Specifications

About this item

LAP3 (leucine aminopeptidase 3), also known as LAPEP or PEPS, is a 519 amino acid protein that localizes to the cytoplasm and belongs to the peptidase M17 family. Existing as a homohexamer, LAP3 uses zinc as a cofactor to catalyze the release of an N-terminal proline from a target peptide and is, therefore, involved in the processing and turnover of intracellular proteins. Multiple isoforms of LAP3 exist due to alternative splicing events. The gene encoding LAP3 maps to human chromosome 4, which houses nearly 6% of the human genome and has the largest gene deserts (regions of the genome with no protein encoding genes) of all of the human chromosomes. Defects in some of the genes located on chromosome 4 are associated with Huntington's disease, Ellis-van Creveld syndrome, methylmalonic acidemia and polycystic kidney disease.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LAP3
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat