Order Entry
Canada
ContactUsLinkComponent
Anti-Synovial sarcoma, X breakpoint 5 Rabbit Polyclonal Antibody (Cy5.5®)
Anti-Synovial sarcoma, X breakpoint 5 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # 10459-768
Supplier:  Bioss
Anti-Synovial sarcoma, X breakpoint 5 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # 10459-768
Supplier:  Bioss
Supplier Number:  BS-7013R-CY5.5

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Synovial sarcoma, X breakpoint 5
  • Antigen symbol:
    SSX5
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5.5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    100 µL
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    6758
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Cat. no.:
    10459-768
  • Supplier No.:
    BS-7013R-CY5.5

Specifications

About this item

The product of this gene belongs to the family of highly homologous synovial sarcoma X (SSX) breakpoint proteins. These proteins may function as transcriptional repressors. They are also capable of eliciting spontaneously humoral and cellular immune responses in cancer patients, and are potentially useful targets in cancer vaccine-based immunotherapy. SSX1, SSX2 and SSX4 genes have been involved in the t(X;18) translocation characteristically found in all synovial sarcomas. This gene appears not to be involved in this type of chromosome translocation. Two transcript variants encoding distinct isoforms have been identified for this gene.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Synovial sarcoma, X breakpoint 5
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human