Anti-TRIM31 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 10433-108
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Tripartite Motif Containing 31
- Antigen symbol:TRIM31
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Gene ID:11074
- Antigen synonyms:RNF|HCGI|C6orf13|Tripartite mot containing protein 31|H.sapiens mRNA for RING protein|Ring finger protein|HCG1|TRI31_HUMAN.
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10433-108
- Supplier No.:BS-6220R-FITC
Specifications
About this item
The TRIM motif includes three zinc binding domains, a RING, a B box type 1 and a B box type 2, and a coiled-coil region. The protein localizes to both the cytoplasm and the nucleus. Its function has not been identified.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TRIM31
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human