Order Entry
Canada
ContactUsLinkComponent
Anti-REG1A Rabbit Polyclonal Antibody (Cy5®)
Anti-REG1A Rabbit Polyclonal Antibody (Cy5®)
Catalog # 10432-374
Supplier:  Bioss
Anti-REG1A Rabbit Polyclonal Antibody (Cy5®)
Catalog # 10432-374
Supplier:  Bioss
Supplier Number:  BS-6183R-CY5

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Regenerating Islet-derived 1 Alpha
  • Antigen symbol:
    REG1A
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    100 µL
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    5967
  • Antigen synonyms:
    PSP|P19|PTP|ICRF|PSPS|PSPS1|REG
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Cat. no.:
    10432-374
  • Supplier No.:
    BS-6183R-CY5

Specifications

About this item

Reg protein is stimulated during the regeneration of pancreatic islets. In human, there are four REG family genes; REG 1 alpha, REG 1 beta, REG-related sequence (RS) and HIP/PAP. These Reg-related proteins are classified into four subfamilies according to their amino-acid sequences, but they share a similar structure and physiological function.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: REG1A
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human