To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Adenovirus 5 E1A
- Antigen symbol:AdV 5 E1A
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Virus
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Protein BS69|Adenovirus 5 E1A-binding protein|MRD30|Bone morphogenetic protein receptor-associated molecule 1|Zinc finger MYND domain-containing protein 11|BRAM1|BS69|ZMYND11
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:180-230/289
- Purification:Purified by Protein A
- Cat. no.:10430-748
- Supplier No.:BS-6136R-FITC
Specifications
About this item
The early region (E1) of the adenovirus genome, responsible for transforming activity, is localized within the left most 11% of the viral genome and consists of two transcriptional units E1A and E1B. E1A is sufficient for partial transformation and immortalization of primary cells. E1A gene products are necessary for normal levels of transcription of the other early regions of the adenovirus genome during productive infection and are able to either activate or repress the transcription of specific cellular genes. E1A forms specific complexes with cellular proteins including p105 causing inhibition of the cell cycle inducing arresting function of p105.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: AdV 5 E1A
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus