Anti-SMYD4 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # 10430-550
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:SET And MYND Domain Containing 4
- Antigen symbol:SMYD4
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rabbit,Human,Rat,Mouse
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Gene ID:114826
- Antigen synonyms:ZMYND21|SMYD4|SMYD4_HUMAN
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10430-550
- Supplier No.:BS-6108R-CY5.5
Specifications
About this item
SMYD4
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SMYD4
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit