To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen name:Chromosome 1 open reading frame 186
- Antigen symbol:C1ORF186
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:440712
- Antigen synonyms:RHEX
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10428-564
- Supplier No.:BS-6023R-FITC
Bone marrow stromal cell antigen 1 (BST1) is a pleiotropic ectoenzyme which belongs to the CD38 family and to the growing number of leukocyte surface molecules known to act independently as both receptors and enzymes. The BST1 molecule displays two distinct domains in its extracellular component. The first is implicated in the enzymic activities of the molecule (it synthesizes cyclic ADP-ribose, a second messenger that elicits calcium release from intracellular stores) and the second domain has adhesion/signalling properties.Bone marrow stromal cell antigen 1 facilitates pre-B-cell growth. The deduced amino acid sequence exhibits 33% similarity with CD38. BST1 expression is enhanced in bone marrow stromal cell lines derived from patients with rheumatoid arthritis. The polyclonal B-cell abnormalities in rheumatoid arthritis may be, at least in part, attributed to BST1 overexpression in the stromal cell population.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: C1ORF186
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat