Order Entry
Canada
Orders LinkContactUsLinkComponent
Anti-EIF4E Rabbit Polyclonal Antibody

Anti-EIF4E Rabbit Polyclonal Antibody

100 µL
Catalog # 10405-244
Supplier:   Bioss
Supplier #:  BS-4979R
Anti-EIF4E Rabbit Polyclonal Antibody
Catalog # 10405-244
Supplier:  Bioss
Anti-EIF4E Rabbit Polyclonal Antibody
Anti-EIF4E Rabbit Polyclonal Antibody

100 µL

Price
$1,053.38
Log in for availability
Each (100µl)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Recognizes and binds the 7-methylguanosine-containing mRNA cap during an early step in the initiation of protein synthesis and facilitates ribosome binding by inducing the unwinding of the mRNAs secondary structures. Component of the CYFIP1-EIF4E-FMR1 complex which binds to the mRNA cap and mediates translational repression. In the CYFIP1-EIF4E-FMR1 complex this subunit mediates the binding to the mRNA cap.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200; IF(ICC)(1:50-200

Type: Primary
Antigen: EIF4E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Documents

Specifications
  • Size:
    100 µL
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Purification:
    Purified by Protein A
  • Antibody type:
    Primary
  • Form:
    liquid
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Storage temperature:
    Store at –20 °C for 12 months
  • Host:
    Rabbit
  • Concentration:
    1 μg/μl
  • Cross adsorption:
    No
  • Shipping temperature:
    4°C
  • Antigen name:
    eukaryotic translation initiation factor 4E
  • Cat. No.:
    10405-244
  • Reactivity:
    Human|Rat|Mouse
  • Gene ID:
    1977
  • Antigen synonyms:
    eIF-4F 25 kDa subunit|EIF4EL1|EIF4E1|eukaryotic translation initiation factor 4E-like 1|eIF-4E|CBP|eIF4E|EIF4F|mRNA cap-binding protein
  • Immunogen:
    170-215/217
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    EIF4E
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.