Order Entry
Canada
ContactUsLinkComponent
 
Anti-NS4B Rabbit Polyclonal Antibody (Cy5®)
  10405-898
 :  Bioss
Anti-NS4B Rabbit Polyclonal Antibody (Cy5®)
  10405-898
 :  Bioss
 :  BS-4857R-CY5

 

  • Antibody type:
    Primary
  • Antigen name:
    Hepatitis C Virus NS4B
  • Antigen symbol:
    NS4B
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Virus
  • Size:
    100 µL
  • Cross adsorption:
    No
  • Form:
    liquid
  • Antigen synonyms:
    HCV|HCV NS4|HepC NS4|HepC
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Cat. no.:
    10405-898
  • Supplier No.:
    BS-4857R-CY5

 

 

HCV is a positive, single-stranded RNA virus in the Flaviviridae family. The genome is approximately 10,000 nucleotides and encodes a single polyprotein of about 3,000 amino acids. The polyprotein is processed by host cell and viral proteases into three major structural proteins and several non-structural protein necessary for viral replication. Several different genotypes of HCV with slightly different genomic sequences have since been identified that correlate with differences in response to treatment with interferon alpha.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NS4B
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus