To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:SHC-transforming Protein 1
- Antigen symbol:SHC1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Gene ID:6464
- Antigen synonyms:SHC|SH2 domain protein C1|FLJ26504|SHC1|p66SHC|Src homology 2 domain-containing-transforming protein C1|SHC-transforming protein 1|p52SHC|p66|SHCA
- Modification:phospho-Tyr317
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10368-722
- Supplier No.:BS-3413R-A488
Specifications
About this item
The SHC gene encodes for a signaling and transforming protein containing Src homology 2 and 3 (SH2 and SH3) domains. The SHC gene encodes 2 widely expressed overlapping proteins of 46 and 52 kD, both containing a C-terminal SH2 domain. Adjacent to the SH2 region is a glycine and proline rich region. These 2 proteins differ in their N terminals. SHC proteins are involved in mitogenic signal transduction and act by coupling growth factor receptors to the RAS signaling pathway. The protein encoded by the SHC1 gene is thought to act as an adaptor in many signal transduction pathways.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SHC1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat