Order Entry
Canada
ContactUsLinkComponent
Anti-IFNGR2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-IFNGR2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 10346-204
Supplier:  Bioss
Anti-IFNGR2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 10346-204
Supplier:  Bioss
Supplier Number:  BS-2710R-HRP
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Interferon gamma receptor 2
  • Antigen symbol:
    IFNGR2
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µL
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    3460
  • Antigen synonyms:
    AF-1|IMD28|IFNGT1|IFGR2
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    270-320/337
  • Purification:
    Purified by Protein A
  • Cat. no.:
    10346-204
  • Supplier No.:
    BS-2710R-HRP

Specifications

About this item

Part of the receptor for interferon gamma. Required for signal transduction. This accessory factor is an integral part of the IFN-gamma signal transduction pathway and is likely to interact with GAF, JAK1, and/or JAK2.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: IFNGR2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat