To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Polyclonal
- Gene ID:4773
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:NFATc2
- Conjugation:Alexa Fluor® 350
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Modification:phospho-Ser326
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10344-018
- Supplier No.:BS-2669R-A350
Specifications
About this item
Nuclear factor of activated T cells (NFAT) is a family of transcription factors implicated in multiple biological processes including cytokine gene expression, cardiac hypertrophy and adipocyte differentiation. NFAT1 (also known as NFATc2 or NFATp) is a member of this family that is regulated by the calcium-dependent phosphatase calcineurin. When calcineurin is activated by calcium it dephosphorylates multiple residues in the regulatory domain of NFAT1, leading to its translocation to the nucleus and activation of its transcriptional activity. Once in the nucleus, NFAT proteins act synergistically with the AP-1 transcription factor complex to regulate the expression of multiple genes. Serine 54 in mouse NFAT1 has been shown to be important in the regulation of its transcriptional activity.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NFATc2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat