Order Entry
ContactUsLinkComponent
Anti-CLEC1B Rabbit Polyclonal Antibody (Cy7®)
Anti-CLEC1B Rabbit Polyclonal Antibody (Cy7®)
Catalog #: 10341-368
Supplier:  Bioss
Anti-CLEC1B Rabbit Polyclonal Antibody (Cy7®)
Catalog #: 10341-368
Supplier:  Bioss
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C-type lectin Domain family 1, member B
  • Clonality:
    Polyclonal
  • Gene ID:
    51266
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Antigen symbol:
    CLEC1B
  • Conjugation:
    Cy7®
  • ImmunoChemistry:
    Yes
  • Size:
    100 µL
  • Cross adsorption:
    No
  • Form:
    liquid
  • Antigen synonyms:
    CLEC2B,
    PRO1384,
    CLEC2,
    1810061I13Rik,
    QDED721
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    1-33/229
  • Purification:
    Purified by Protein A
  • Cat. no.:
    10341-368
  • Supplier No.:
    BS-2543R-CY7

Specifications

About this item

Natural killer (NK) cells express multiple C-type (calcium-dependent) lectin-like receptors, that interact with major histocompatibility complex class I molecules and either inhibit or activate cytotoxicity and cytokine secretion. CLEC1B (also known as CLEC-2) is a C-type lectin-like receptor expressed in myeloid cells and NK cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CLEC1B
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought