Order Entry
Canada
ContactUsLinkComponent
Anti-CLEC1A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-CLEC1A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 10341-298
Supplier:  Bioss
Anti-CLEC1A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 10341-298
Supplier:  Bioss
Supplier Number:  BS-2542R-HRP

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C-type lectin Domain family 1, member A
  • Antigen symbol:
    CLEC1A
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µL
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    51267
  • Antigen synonyms:
    CLEC-1|CLEC1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    160-210/280
  • Purification:
    Purified by Protein A
  • Cat. no.:
    10341-298
  • Supplier No.:
    BS-2542R-HRP

Specifications

About this item

This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded protein may play a role in regulating dendritic cell function. Alternative splice variants have been described but their biological nature has not been determined. This gene is closely linked to other CTL/CTLD superfamily members on chromosome 12p13 in the natural killer gene complex region. [provided by RefSeq]

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: CLEC1A
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: