Order Entry
ContactUsLinkComponent
Anti-TNFSF18 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog #: 10339-988
Supplier:  Bioss
Price
$1,281.54
Log in for availability
Unit of Measure Each (100µl)
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Tumor Necrosis Factor Superfamily Member 18
  • Clonality:
    Polyclonal
  • Gene ID:
    8995
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
    Rat
    Mouse
  • Antigen symbol:
    TNFSF18
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ImmunoChemistry:
    Yes
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    liquid
  • Antigen synonyms:
    hGITRL
    Activation-inducible TNF-related ligand
    TL6
    Activation-inducible TNF-related Ligand
    AITRL
    UNQ149/PRO175
    GITRL
    Tumor necrosis factor ligand superfamily member 18
    TNFSF18
    Glucocorticoid-induced TNF Receptor Ligand
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    150-180/199
  • Purification:
    Purified by Protein A
  • Cat. no.:
    10339-988
  • Size:
    100 µL

Specifications

About this item