Anti-duck hepatitis A virus 1 Genome polyprotein Rabbit Polyclonal Antibody (Cy7®)
Catalog # 10326-374
Supplier: Bioss
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:duck hepatitis A virus 1 Genome polyprotein
- Antigen symbol:duck hepatitis A virus 1 Genome polyprotein
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Virus
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:880-910/2249
- Purification:Purified by Protein A
- Cat. no.:10326-374
- Supplier No.:BS-2197R-CY7
Specifications
About this item
duck hepatitis A virus genome polyprotein
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: duck hepatitis A virus 1 Genome polyprotein
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus