HAEMO-SOL® Enzyme Active, Detergent
Supplier: Haemo-Sol
Certificates
About this item
Concentrated enzymatic detergent for hard to remove protein wastes. Breaks down dried, heavy or hardened soils for easy removal after soaking.
Mix 1/2 ounce to 1 gallon of warm water. Soak instruments 20 minutes or more if heavliy soiled. Use a soft brush if necessary. 5 lb makes 160 gallons of solution.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together