Orders LinkContactUsLinkComponent
Cryogenic Printer Labels, Brady
Cryogenic Printer Labels, Brady
Cryogenic Printer Labels, Brady
Supplier:  Avantor
Certificates
Cryogenic Printer Labels, Brady
Cryogenic Printer Labels, Brady
6 Options Available Below
Product Details & Documents
Cryogenic labels for tubes, vials, and canes.
Chemical and stain resistant labels for slides and cassettes:
Temperatures from –80 to 121 °C (–112 to 250 °F)
B-488 Slide label with moderate chemical resistance
B-498 Tissue cassette label for re-identification of processed paraffin blocks.
General labels for straws, petri dishes, bottles, flasks, microplates, bags, and more:
B-499 For cryo boxes, freezer racks, petri dishes, bottles, flasks, microplates, and bags, working temperatures from –196 to 121 °C (–321 to 250 °F)
B-595 General purpose label for drawers, shelves, and floor.
Product Family Options
B-461 Ultra-Thin Label with Clear Tail for Tubes
Product Information
  • Material
    Color
    Width
    Dimensions
    Length
    Availability
    Price
  • CA80090-426
    Labxpert labels B-461 MAT´L
    Labxpert labels B-461 MAT´L
    Polyester
    Black on white
    9.5×482.6 mm (0.38×19")
    Log in for availability
    Log in for availability
    Each
    Each - $110.06
    $110.06
    Each
       $110.06
  • CA80090-424
    Labxpert labels B-461 MAT´L
    Labxpert labels B-461 MAT´L
    Polyester
    Black on white
    31.8×482.6 mm (1.25×19")
    Log in for availability
    Log in for availability
    Each
    Each - $96.99
    $96.99
    Each
       $96.99
  • 490006-484
    Cryogenic printer labels
    Cryogenic printer labels
    Polyester
    Black on white
    127×9.1 mm (5×0.36")
    Discontinued
  • 490006-478
    Cryogenic printer labels
    Cryogenic printer labels
    Polyester
    Black on white
    25.4×482.6 mm (1×19")
    Discontinued
B-499 For Cryo Boxes, Freezer Racks, Petri Dishes, Bottles, Flasks, Microplates, and Bags
Product Information
  • Material
    Color
    Width
    Dimensions
    Length
    Availability
    Price
  • CA80090-398
    Labxpert labels B-499 MAT´L
    Labxpert labels B-499 MAT´L
    Nylon Cloth
    Black on white
    38.1×31.8 mm (1.5×1.25")
    Log in for availability
    Log in for availability
    Each
    Each - $163.42
    $163.42
    Each
       $163.42
  • 490006-458
    Cryo printer labels 9.5 mm
    Cryo printer labels 9.5 mm
    Nylon Cloth
    Black on white
    Discontinued
  • 490006-438
    Cryogenic printer labels
    Cryogenic printer labels
    Nylon Cloth
    Black on white
    127×12.7 mm (5×0.5")
    Discontinued
  • 490006-468
    Cryo printer labels 12.7 mm
    Cryo printer labels 12.7 mm
    Nylon Cloth
    Black on white
    Discontinued
  • 490006-474
    Cryo printer labels 12.7 mm
    Cryo printer labels 12.7 mm
    Nylon Cloth
    Black on yellow
    Discontinued
  • 490006-446
    Cryogenic printer labels
    Cryogenic printer labels
    Nylon Cloth
    Black on white
    19.1×38.1 mm (0.75×1.5")
    Discontinued
  • 490006-452
    Cryogenic printer labels
    Cryogenic printer labels
    Nylon Cloth
    Black on white
    48.3×25.4 mm (1.9×1")
    Discontinued
B-595 General Purpose Label for Drawers, Shelves, and Floor
Product Information
  • Material
    Color
    Width
    Dimensions
    Length
    Availability
    Price
  • CA89205-172
    Labxpert labels B-595 MAT´L
    Labxpert labels B-595 MAT´L
    Vinyl
    Black on white
    127×482.6 mm (5×19")
    Log in for availability
    Log in for availability
    Each
    Each - $70.99
    $70.99
    Each
       $70.99
  • CA89205-182
    Labxpert labels B-595 MAT´L
    Labxpert labels B-595 MAT´L
    Vinyl
    Black on white
    25.4×482.6 mm (1×19")
    Log in for availability
    Log in for availability
    Each
    Each - $98.99
    $98.99
    Each
       $98.99
  • Discontinued
  • Discontinued
  • CA89205-212
    Labxpert labels B-595 MAT´L
    Labxpert labels B-595 MAT´L
    Vinyl
    Black on white
    38.1×482.6 mm (1.5×19")
    Log in for availability
    Log in for availability
    Each
    Each - $117.99
    $117.99
    Each
       $117.99
  • 490006-488
    Cryogenic printer labels
    Cryogenic printer labels
    Vinyl
    White on blue
    127×482.6 mm (5×19")
    Discontinued
  • 490006-490
    Cryogenic printer labels
    Cryogenic printer labels
    Vinyl
    Black on white
    127×482.6 mm (5×19")
    Discontinued