Order Entry
Canada
Orders LinkContactUsLinkComponent
H-Tyr(Tbu)-2-Chlorotrityl

H-Tyr(Tbu)-2-Chlorotrityl

Supplier:   Bachem Americas
H-Tyr(Tbu)-2-Chlorotrityl
Supplier:  Bachem Americas
Certificates

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • D-2890.0001BA
  • D-2890.0005BA
  • Protein/peptide name:
  • H-Tyr(Tbu)-2-Chlorotrityl
  • H-Tyr(Tbu)-2-Chlorotrityl
  • MDL No.:
  • MFCD00801420
  • MFCD00801420
  • Sequence:
  • H-Tyr(tBu)-2-chlorotrityl resin (200-400 mesh) (Low Substitution)
  • H-Tyr(tBu)-2-chlorotrityl resin (200-400 mesh) (Low Substitution)
  • Storage conditions:
  • –20 ± 5 °C
  • –20 ± 5 °C

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $309.91
    $309.91
    Each (1g)
       $309.91
    Minimum order quantity is 1

    Supplier Number: 4068311.0001
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    D-2890.0001BA
    Protein/peptide name
    H-Tyr(Tbu)-2-Chlorotrityl
    MDL No.
    MFCD00801420
    Sequence
    H-Tyr(tBu)-2-chlorotrityl resin (200-400 mesh) (Low Substitution)
    Storage conditions
    –20 ± 5 °C
  • 5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $1,247.83
    $1,247.83
    Each (5g)
       $1,247.83
    Minimum order quantity is 1

    Supplier Number: 4068311.0005
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    D-2890.0005BA
    Protein/peptide name
    H-Tyr(Tbu)-2-Chlorotrityl
    MDL No.
    MFCD00801420
    Sequence
    H-Tyr(tBu)-2-chlorotrityl resin (200-400 mesh) (Low Substitution)
    Storage conditions
    –20 ± 5 °C