Ward's® Introduction to Agarose Gel Electrophoresis Lab Activity
Catalog # 470024-004
Supplier: Avantor
Specifications
- Description:Intro to Agarose Gel Electrophoresis Kit
- Environmentally Preferable:
- No. of Lab Groups:6
- Cat. no.:470024-004
- Supplier No.:WARD470024-004
Specifications
About this item
This fast and easy activity acquaints students with electrophoresis using inexpensive dyes.
- Kit employs colorful dyes to simulate DNA
- Materials for six lab groups
- Time required: approximately 30 minutes
- Teacher's guide included
This lab activity helps students become accustomed to the electrophoresis process before they work with DNA. They use specially formulated dyes, which charge and separate based on their molecular size when electrophoresed, on an agarose gel; students then determine the missing dye component in a mixture.
Accessories information: An electrophoresis chamber, power supply, and 10 μL micropipets, required, are available separately.
Ordering information: An electrophoresis chamber, power supply, and 10μL micropipets, required, are available separately.
Delivery: This kit comes with enough material to run up to six gels including prepared agarose that needs only to be heated and poured as well as a teacher’s guide and student copy master.