Orders LinkContactUsLinkComponent
Bucket, Rectangular, Vileda Professional-FHP
Bucket, Rectangular, Vileda Professional-FHP
Bucket, Rectangular, Vileda Professional-FHP
Certificates
Bucket, Rectangular, Vileda Professional-FHP
Bucket, Rectangular, Vileda Professional-FHP
2 Options Available Below
Product Details & Documents
High quality polypropylene buckets with 6 L capacity. Can be used individually and also on the CE Bucket Trolley or CE Pre-Prepared Trolley.
  • Color coding clips included to allow differentiation in colors of buckets (red, blue, green, yellow)
  • Autoclavable
Ordering information: Lid sold separately.
Product Family Options
Product Information
  • Capacity
    Dimensions
    Availability
    Price
  • 10153-180
    Bucket, without lid
    Bucket, without lid
    6 L
    11×8×7"
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $86.30
    $86.30
    Log in for availability
    Case of 10
    Case of 10 - $863.00
    $863.00
    Each
       $86.30
    Case of 10
       $863.00
    Supplier Number: 120942
  • 10153-182
    Lid for 6 L Bucket, autoclavable
    Lid for 6 L Bucket, autoclavable
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $23.30
    $23.30
    Log in for availability
    Case of 12
    Case of 12 - $279.60
    $279.60
    Each
       $23.30
    Case of 12
       $279.60
    Supplier Number: 125948