Specifications
- Description:Littauer Scissor
- Material:Stainless Steel
- Cat. no.:89023-166
- Length:89 mm (3 1/2")
- Supplier No.:21-346
- Blade Type:Smooth, Curved, Blunt/Blunt
Specifications
About this item
Econo™ Littauer Suture Scissors are a disposable, floor-grade instrument used for suture removal.
These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. Littauer suture scissors have larger, less delicate tips than Spencer Stitch Scissors, and are often used for removing larger sutures. They should not be used for cutting tissue.
Econo™ instruments are Sklar's floor-grade, disposable instruments.