ContactUsLinkComponent
Oscillating Reactions: The Traffic Light
  470219-088
undefined
Oscillating Reactions: The Traffic Light
  470219-088
 :  IS7024
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Oscillating Reactions - The Traffic Light
  • Environmentally Preferable:
  • Cat. no.:
    470219-088
  • Supplier No.:
    IS7024
  • No. of Uses:
    5
Are chemical reactions reversible?
  • Redox reaction illustrates chemical equilibrium
  • Easy way for students to see chemical reactions taking place
  • Cost and Time Efficient
  • Meets National Science Education Standards for grades 5-12
  • Time Requirement: 30 minutes
Oscillating reactions demonstrate chemical equilibrium. A reversible color change is imparted on the chromosphore indigo carmine. Energy (swirling or shaking) is used to drive the reaction. The prepared solution may be used for up to an hour, at which time, a fresh solution should be prepared.
 : Materials Included are Glucose, Sodium hydroxide, and Indigo carmine QuickPrep. Materials Needed But Not Included are distilled or deionized water, balance, and erlenmeyer flask, graduated cylinders