Order Entry
Canada
ContactUsLinkComponent
H-Tyr(Tbu)-2-Chlorotrityl Res
H-Tyr(Tbu)-2-Chlorotrityl Res
Supplier:  Bachem Americas
Certificates
H-Tyr(Tbu)-2-Chlorotrityl Res
Supplier:  Bachem Americas

Specifications

  • Storage conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD00801420
  • Protein/peptide name:
    H-Tyr(Tbu)-2-Chlorotrityl Res
  • Sequence:
    H-Tyr(tBu)-2-chlorotrityl resin (200-400 mesh)

Specifications

Frequently Bought Together