Order Entry
Canada
ContactUsLinkComponent
Volumetric Saybolt Viscosity Flask, Class A, DWK Life Sciences
Volumetric Saybolt Viscosity Flask, Class A, DWK Life Sciences
Catalog # 10000-518
undefined
Volumetric Saybolt Viscosity Flask, Class A, DWK Life Sciences
Catalog # 10000-518
Supplier Number:  28126-60

Specifications

  • Capacity:
    60 ml
  • Environmentally Preferable:
  • Cat. no.:
    10000-518
  • Tolerance:
    ±0.10 ml
  • Supplier No.:
    28126-60

Specifications

About this item

The Ramsbottom Coking Bulb is used in the determination of carbon residue in petroleum products.

Ref: ASTM D524.