CORIO™ C Open Heating Bath Circulators, JULABO
Supplier: JULABO
Certificates
About this item
CORIO™ C heating circulator combinations with transparent PC or stainless steel bath tanks for internal temperature applications from +20 to +100 °C
Products include a quick connect clamp for positioning CORIO C on tank rim. Polycarbonate bath tanks feature volume graduations in L and Qt. Stainless steel tanks include a drain tap (except B5).
To adapt to your individual application we offer a comprehensive range of accessories.
Certifications: CE marked.
Ordering information: Includes CORIO C immersion circulator, bath clamp and bath tank.