KLEENGUARD® A20 Breathable Particle Protection Frocks, Kimberly-Clark
Supplier: Microflex
Certificates
About this item
KLEENGUARD® A20 SMMMS frocks offer superior particle holdout coupled with unmatched comfort attributes, providing workers protection without sacrificing comfort against non-hazardous and hazardous dust
- Superior particle holdout coupled with unmatched comfort attributes
- 5 layers of stacked barrier protection (SMMMS) against hazardous particles
Lightweight and breathable. Serged seams, five-snap closure, elastic wrists, below knee length, and mandarin collar.
The A20 fabric has five layers of stacked barrier protection (SMMMS) against hazardous particles.
Certifications: NFPA 99 for Antistatic Materials. Eligible for recycling with KIMBERLY-CLARK PROFESSIONAL®
Specifications
- Catalog No:
- CA10815-980
- CA10815-982
- CA10815-984
- CA10815-986
- Fabric:
- SMMMS
- SMMMS
- SMMMS
- SMMMS
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Pockets:
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory