Oxytocin ELISA kit, Enzo Life Sciences
10014-684
:
- Sensitivity:15 pg/mL (15.6 - 1,000 pg/mL)
- Cat. no.:10014-684
- Supplier No.:900-153A-0001
- No. of Tests:96 wells
The most sensitive, specific and flexible alternative to more costly and time-consuming methods for oxytocin detection.
For the quantitative determination of oxytocin in culture supernatants, milk, plasma, and serum from any species. Cited sample types include cerebral spinal fluid, saliva, tissue and urine.
: Contains GxR IgG microtiter plate, conjugate, antibody, assay buffer, wash buffer concentrate, standard, pNpp substrate, stop solution. Alternative size available: ADI-901-153A (5 x 96 wells).