About this item
FastPROBE™ qPCR No-ROX Master Mix is ready-to-use cocktail for the amplification and quantitation of DNA on real-time instruments that do not incorporate a ROX passive reference dye
An integral component, the Classic++ Hot Start Taq DNA Polymerase is compatible with all fluorogenic probes including hydrolysis and displacement probes such as TaqMan®, Scorpions® and Molecular Beacons probes. Novel hot start technology prevents primer dimer formation and non-specific amplification resulting in higher yields and specificity. Advanced buffer chemistry means little to no protocol optimization, regardless of template.
FastPROBE qPCR Master Mixes are suitable for both standard and fast-cycling real-time PCR applications, providing improved signal to noise ratios, rapid extension rates for early Ct values, and enhanced sensitivity. Use for low copy number detection, microarray validation, SNP genotyping and multiplexing - Please refer to our Instrument Compatibility Guide to ensure you have the correct ROX concentration for your system.
Specifications
- Application:qPCR probe