Baseline® Three Point Descriminator
Catalog # 470119-640
Supplier: American 3B Scientific
Specifications
- Description:Three Point Descriminator
- Environmentally Preferable:
- Cat. no.:470119-640
- Supplier No.:W54068
Specifications
About this item
Accurately test two point sensitivity.
- Sliding point measured in mm
- Switch between one and two point touch easily
Evaluate cutaneous sensitivity and touch threshold. Plastic points minimize influence of temperature. Two point discriminator has third point that permits alternating between single and double point stimulation without changing setting, measures up to 14 cm discrimination.