About this item
CYBRFast qPCR Hi-ROX Master Mix is a ready-to-use cocktail for the amplification and detection of DNA on real-time instruments that support normalization with a standard ROX passive reference dye at a final concentration of 500 nM
CYBRFast GREEN DNA intercalating dye does not inhibit PCR, resulting in better fluorescent signals than other dyes, and is optimized for use with CYBRFast Hot Start Taq DNA Polymerase. Novel hot start technology prevents primer dimer formation and non-specific amplification resulting in higher yields and specificity. Advanced buffer chemistry means little to no protocol optimization, regardless of template.
CYBRFast qPCR Master Mixes in real-time applications for improved signal to noise ratios, rapid extension rates for early Ct values, and enhanced sensitivity.This mix is ideal for high-throughput qPCR, and can be used with a variety of complex targets including genomic, cDNA and viral sequences - Please refer to our Instrument Compatibility Guide to ensure you have the correct ROX concentration for your system.
It reliably delivers enhanced fluorescence, speed, sensitivity and linearity.
Specifications
- Application:qPCR dye